Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20605 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033148.1 | 18 | 3108 | |||
GenBank mRNA | AK164339.1 | - | 1 | 315 |
Gene Symbol: Lrch4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131167.1 | - | 3268 |