Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330024D21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015582.2 | 3 | 314 | |||
NR_110363.1 | 3 | 314 | ||||
GenBank mRNA | AK049146.1 | - | 3 | 309 | ||
AK049327.1 | - | 3 | 315 | |||
AK169674.1 | - | 3 | 312 | |||
AK169807.1 | - | 3 | 312 |