Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pea15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC132661.1 | - | 22 | 40130938 | ||
BC132663.1 | - | 22 | 40130940 |