Search
Search

DPY19L2P4| Gene Symbol: DPY19L2P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003551.1 | 3 | 1149 | |||
| GenBank mRNA | BC031277.1 | - | 3 | 1106 | ||
| BC047471.1 | - | 3 | 1097 | |||
| BX537815.1 | - | 3 | 1149 | |||
| DQ287934.1 | - | 3 | 1149 | |||