Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CMAHP SIRNA Assays ›
          Gene Symbol:
          CMAHP
          Gene Name:
          cytidine monophospho-N-acetylneuraminic acid hydroxylase, pseudogene
          Gene Aliases:
          CMAH, CSAH
          Chromosome Location:
          Chr.6: 25081067 - 25138392 on Build GRCh38
          UniGene ID:
          Hs.484918
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258472

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CMAHP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002174.2 1 17
          NR_027626.1 2 336
          GenBank mRNA AF074480.1 - 2 37
          AK303297.1 - 2 293
          BC032500.1 - 2 229
          DA976765.1 - 2 337
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX417923.2 - 2 197 Chr. 6 - 25108620 - 25218530 [-] on Build GRCh38
          GenBankmRNA DB283113.1 - 2 273 Chr. 6 - 25115014 - 25138329 [-] on Build GRCh38
          GenBankmRNA AL583055.3 - 2 74 Chr. 6 - 25106377 - 25138582 [-] on Build GRCh38
          GenBankmRNA DA845166.1 - 2 273 Chr. 6 - 25114999 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB273230.1 - 2 273 Chr. 6 - 25115001 - 25138329 [-] on Build GRCh38
          GenBankmRNA CR979796.1 - 1 33 Chr. 6 - 25106441 - 25137825 [-] on Build GRCh38
          GenBankmRNA DB275975.1 - 2 274 Chr. 6 - 25114996 - 25138330 [-] on Build GRCh38
          GenBankmRNA DA984771.1 - 2 273 Chr. 6 - 25115010 - 25138329 [-] on Build GRCh38
          GenBankmRNA DN996058.1 - 2 205 Chr. 6 - 25113693 - 25218539 [-] on Build GRCh38
          GenBankmRNA DB114213.1 - 2 271 Chr. 6 - 25115004 - 25138326 [-] on Build GRCh38
          GenBankmRNA DB281666.1 - 2 273 Chr. 6 - 25115000 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA640388.1 - 2 274 Chr. 6 - 25114989 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA945677.1 - 2 277 Chr. 6 - 25115014 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB263995.1 - 2 239 Chr. 6 - 25114929 - 25138295 [-] on Build GRCh38
          GenBankmRNA DA357127.1 - 2 39 Chr. 6 - 25110083 - 25138095 [-] on Build GRCh38
          GenBankmRNA DA974280.1 - 2 278 Chr. 6 - 25114989 - 25138329 [-] on Build GRCh38
          GenBankmRNA CB990369.1 - 355 Chr. - - [-] on Build
          GenBankmRNA AV757536.2 - 2 347 Chr. 6 - 25113619 - 25166557 [-] on Build GRCh38
          GenBankmRNA DA853213.1 - 2 224 Chr. 6 - 25113740 - 25218555 [-] on Build GRCh38
          GenBankmRNA DA814648.1 - 2 273 Chr. 6 - 25115012 - 25138329 [-] on Build GRCh38
          GenBankmRNA AL558196.3 - 2 263 Chr. 6 - 25108620 - 25166535 [-] on Build GRCh38
          GenBankmRNA DB152436.1 - 2 273 Chr. 6 - 25114956 - 25138329 [-] on Build GRCh38
          GenBankmRNA BI835680.1 - 2 230 Chr. 6 - 25110057 - 25138285 [-] on Build GRCh38
          GenBankmRNA DA965960.1 - 2 273 Chr. 6 - 25114996 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB291396.1 - 2 273 Chr. 6 - 25114987 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB269255.1 - 2 228 Chr. 6 - 25109546 - 25138284 [-] on Build GRCh38
          GenBankmRNA DA619313.1 - 2 273 Chr. 6 - 25113749 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB103158.1 - 2 278 Chr. 6 - 25114987 - 25218611 [-] on Build GRCh38
          GenBankmRNA DB150549.1 - 2 267 Chr. 6 - 25114989 - 25138323 [-] on Build GRCh38
          GenBankmRNA CB269549.1 - 2 210 Chr. 6 - 25113674 - 25138266 [-] on Build GRCh38
          GenBankmRNA DC361891.1 - 2 262 Chr. 6 - 25115024 - 25166534 [-] on Build GRCh38
          GenBankmRNA DB118779.1 - 2 232 Chr. 6 - 25114995 - 25138287 [-] on Build GRCh38
          GenBankmRNA DB016444.1 - 2 228 Chr. 6 - 25113740 - 25138284 [-] on Build GRCh38
          GenBankmRNA CF994240.1 - 2 218 Chr. 6 - 25109555 - 25218524 [-] on Build GRCh38
          GenBankmRNA BM926468.1 - 273 Chr. - - [-] on Build
          GenBankmRNA DB100822.1 - 2 273 Chr. 6 - 25113595 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA422585.1 - 2 273 Chr. 6 - 25114997 - 25138329 [-] on Build GRCh38
          GenBankmRNA CB988240.1 - 2 337 Chr. 6 - 25113663 - 25218644 [-] on Build GRCh38
          GenBankmRNA DA953243.1 - 2 277 Chr. 6 - 25115007 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA987420.1 - 2 273 Chr. 6 - 25114991 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA933476.1 - 2 273 Chr. 6 - 25114991 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB049596.1 - 2 274 Chr. 6 - 25114996 - 25138329 [-] on Build GRCh38
          GenBankmRNA CR975817.1 - 1 34 Chr. 6 - 25108580 - 25137825 [-] on Build GRCh38
          GenBankmRNA DB137826.1 - 2 275 Chr. 6 - 25114994 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA945691.1 - 2 274 Chr. 6 - 25115009 - 25138328 [-] on Build GRCh38
          GenBankmRNA DA639718.1 - 2 277 Chr. 6 - 25115024 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA535830.1 - 3 243 Chr. 6 - 25113763 - 25167458 [-] on Build GRCh38
          GenBankmRNA DB251587.1 - 2 273 Chr. 6 - 25115026 - 25138329 [-] on Build GRCh38
          GenBankmRNA CB961020.1 - 4 360 Chr. 6 - 25113650 - 25218507 [-] on Build GRCh38
          GenBankmRNA DC406723.1 - 2 265 Chr. 6 - 25114989 - 25166537 [-] on Build GRCh38
          GenBankmRNA DB279857.1 - 2 273 Chr. 6 - 25115026 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB213851.1 - 2 274 Chr. 6 - 25115016 - 25138330 [-] on Build GRCh38
          GenBankmRNA DA951980.1 - 2 178 Chr. 6 - 25114909 - 25138230 [-] on Build GRCh38
          GenBankmRNA DC392793.1 - 2 148 Chr. 6 - 25113678 - 25166420 [-] on Build GRCh38
          GenBankmRNA DB059853.1 - 2 273 Chr. 6 - 25115023 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA622241.1 - 2 273 Chr. 6 - 25115017 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA466388.1 - 2 277 Chr. 6 - 25115006 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB261379.1 - 2 273 Chr. 6 - 25114991 - 25138328 [-] on Build GRCh38
          GenBankmRNA DB200971.1 - 2 240 Chr. 6 - 25113735 - 25138295 [-] on Build GRCh38
          GenBankmRNA DA921665.1 - 2 273 Chr. 6 - 25114987 - 25138329 [-] on Build GRCh38
          GenBankmRNA CD638396.1 - 2 95 Chr. 6 - 25110069 - 25138148 [-] on Build GRCh38
          GenBankmRNA DA627865.1 - 2 228 Chr. 6 - 25114989 - 25138284 [-] on Build GRCh38
          GenBankmRNA DB029512.1 - 2 273 Chr. 6 - 25114994 - 25138329 [-] on Build GRCh38
          GenBankmRNA CR982714.1 - 2 72 Chr. 6 - 25113646 - 25138075 [-] on Build GRCh38
          GenBankmRNA DA014839.1 - 3 273 Chr. 6 - 25115010 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA983830.1 - 2 272 Chr. 6 - 25115002 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA909593.1 - 2 266 Chr. 6 - 25113764 - 25138315 [-] on Build GRCh38
          GenBankmRNA DA636732.1 - 2 273 Chr. 6 - 25114987 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB276249.1 - 2 228 Chr. 6 - 25113766 - 25138284 [-] on Build GRCh38
          GenBankmRNA DA393082.1 - 2 273 Chr. 6 - 25113758 - 25138329 [-] on Build GRCh38
          GenBankmRNA CD640511.1 - 2 49 Chr. 6 - 25110025 - 25138102 [-] on Build GRCh38
          GenBankmRNA DB284942.1 - 2 273 Chr. 6 - 25115013 - 25138329 [-] on Build GRCh38
          GenBankmRNA DB126765.1 - 3 214 Chr. 6 - 25113662 - 25167430 [-] on Build GRCh38
          GenBankmRNA DA681557.1 - 2 228 Chr. 6 - 25113726 - 25138284 [-] on Build GRCh38
          GenBankmRNA CD520373.1 - 2 45 Chr. 6 - 25109965 - 25138101 [-] on Build GRCh38
          GenBankmRNA DB066123.1 - 2 265 Chr. 6 - 25115006 - 25138321 [-] on Build GRCh38
          GenBankmRNA CF994709.1 - 4 360 Chr. 6 - 25115002 - 25218507 [-] on Build GRCh38
          GenBankmRNA DB258372.1 - 2 273 Chr. 6 - 25114997 - 25138329 [-] on Build GRCh38
          GenBankmRNA DA860408.1 - 3 225 Chr. 6 - 25114996 - 25218498 [-] on Build GRCh38
          GenBankmRNA DA981207.1 - 2 262 Chr. 6 - 25113755 - 25138317 [-] on Build GRCh38
          GenBankmRNA DC423527.1 - 2 265 Chr. 6 - 25115012 - 25166537 [-] on Build GRCh38
          GenBankmRNA DB199970.1 - 2 220 Chr. 6 - 25113738 - 25218553 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydroxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of Wnt signaling pathway
          CMP-N-acetylneuraminate monooxygenase activity
          2 iron, 2 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0