Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RCC1 SIRNA Assays ›
          Gene Symbol:
          RCC1 SNHG3
          Gene Name:
          regulator of chromosome condensation 1
          small nucleolar RNA host gene 3
          Gene Aliases:
          CHC1, RCC1-I, SNHG3-RCC1
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          Chromosome Location:
          Chr.1: 28505943 - 28539196 on Build GRCh38
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          UniGene ID:
          Hs.469723
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n258485

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BF667678.1 - 3 541
          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 545
          NR_036473.1
          4 653
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 445
          AJ006835.1 - LIT1315
          U17HG
          3 445
          AK092726.1 -
          2 2178
          AK311577.1 -
          1 197
          BC006790.1 -
          3 427
          BC012122.1 -
          3 433
          BC017334.2 -
          3 420
          DA044266.1 -
          3 504
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA820174.1 - 3 442 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 422 Chr. - - [-] on Build
          GenBankmRNA DA998284.1 - 3 453 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA CN366542.1 - 4 381 Chr. 1 - 28506055 - 28509581 [+] on Build GRCh38
          GenBankmRNA DA955189.1 - 3 447 Chr. 1 - 28506043 - 28509267 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 379 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA DB113121.1 - 3 445 Chr. 1 - 28506044 - 28509329 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 445 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA HY182836.1 - 3 443 Chr. 1 - 28506045 - 28509242 [+] on Build GRCh38
          GenBankmRNA DA928567.1 - 3 442 Chr. 1 - 28506047 - 28509337 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 430 Chr. - - [-] on Build
          GenBankmRNA DW417474.1 - 90 Chr. - - [-] on Build
          GenBankmRNA CB985655.1 - 3 448 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 440 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA DA969742.1 - 3 449 Chr. 1 - 28506045 - 28509315 [+] on Build GRCh38
          GenBankmRNA DA968918.1 - 3 444 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM802102.1 - 432 Chr. - - [-] on Build
          GenBankmRNA BM790281.1 - 3 432 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA AV763089.1 - 2 438 Chr. 1 - 28508127 - 28509528 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 463 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 444 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA DA089660.1 - 3 505 Chr. 1 - 28505984 - 28509274 [+] on Build GRCh38
          GenBankmRNA HY176129.1 - 3 443 Chr. 1 - 28506045 - 28509283 [+] on Build GRCh38
          GenBankmRNA EB387088.1 - 1 106 Chr. 1 - 28509094 - 28509634 [+] on Build GRCh38
          GenBankmRNA DA704998.1 - 3 444 Chr. 1 - 28506044 - 28509252 [+] on Build GRCh38
          GenBankmRNA AV689452.1 - 3 475 Chr. 1 - 28506009 - 28509366 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 444 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 444 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA BE867363.1 - 2 419 Chr. 1 - 28508131 - 28509511 [+] on Build GRCh38
          GenBankmRNA DB024065.1 - 1 197 Chr. 1 - 28509002 - 28509575 [+] on Build GRCh38
          GenBankmRNA DA918003.1 - 3 445 Chr. 1 - 28506044 - 28509337 [+] on Build GRCh38
          GenBankmRNA DA817644.1 - 3 445 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA847021.1 - 3 445 Chr. 1 - 28506044 - 28509325 [+] on Build GRCh38
          GenBankmRNA HY177570.1 - 3 443 Chr. 1 - 28506045 - 28509225 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 444 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 417 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 427 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA961704.1 - 3 445 Chr. 1 - 28506044 - 28509303 [+] on Build GRCh38
          GenBankmRNA HY179875.1 - 3 442 Chr. 1 - 28506045 - 28509263 [+] on Build GRCh38
          GenBankmRNA DA922155.1 - 3 445 Chr. 1 - 28506044 - 28509320 [+] on Build GRCh38
          GenBankmRNA AV764560.2 - 3 435 Chr. 1 - 28506051 - 28509412 [+] on Build GRCh38
          GenBankmRNA DB157161.1 - 3 446 Chr. 1 - 28506043 - 28509332 [+] on Build GRCh38
          GenBankmRNA DA644956.1 - 3 445 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA BM479709.1 - 3 430 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 398 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA HY173512.1 - 3 444 Chr. 1 - 28506045 - 28509232 [+] on Build GRCh38
          GenBankmRNA DA925005.1 - 3 444 Chr. 1 - 28506044 - 28509319 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 422 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 432 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 421 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA DA936462.1 - 3 444 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DB112428.1 - 3 445 Chr. 1 - 28506044 - 28509353 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 416 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 414 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA DA952260.1 - 3 445 Chr. 1 - 28506044 - 28509313 [+] on Build GRCh38
          GenBankmRNA BE841288.1 - 1 130 Chr. 1 - 28509097 - 28509254 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 323 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 448 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 428 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA DA565956.1 - 3 444 Chr. 1 - 28506044 - 28509324 [+] on Build GRCh38
          GenBankmRNA HY084156.1 - 3 443 Chr. 1 - 28506045 - 28509217 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 444 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA BG474710.1 - 3 435 Chr. 1 - 28506055 - 28509618 [+] on Build GRCh38
          GenBankmRNA CB157773.1 - 3 433 Chr. 1 - 28506055 - 28509221 [+] on Build GRCh38
          GenBankmRNA DB262011.1 - 3 445 Chr. 1 - 28506044 - 28509330 [+] on Build GRCh38
          GenBankmRNA DB271395.1 - 3 445 Chr. 1 - 28506044 - 28509295 [+] on Build GRCh38
          GenBankmRNA DA943290.1 - 3 446 Chr. 1 - 28506043 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY089919.1 - 3 443 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 499 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA HY179907.1 - 3 443 Chr. 1 - 28506045 - 28509261 [+] on Build GRCh38
          GenBankmRNA DA510712.1 - 3 445 Chr. 1 - 28506044 - 28509216 [+] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 445 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA BF982554.1 - 2 390 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA DA787816.1 - 3 444 Chr. 1 - 28506044 - 28509270 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 435 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA BG023899.1 - 413 Chr. - - [-] on Build
          GenBankmRNA DA902617.1 - 3 444 Chr. 1 - 28506045 - 28509297 [+] on Build GRCh38
          GenBankmRNA HY176989.1 - 3 442 Chr. 1 - 28506045 - 28509229 [+] on Build GRCh38
          GenBankmRNA HY175139.1 - 3 443 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 442 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA CD652956.1 - 1 72 Chr. 1 - 28509129 - 28509633 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 413 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 488 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA CN366539.1 - 3 420 Chr. 1 - 28506068 - 28509242 [+] on Build GRCh38
          GenBankmRNA HY182945.1 - 3 443 Chr. 1 - 28506045 - 28509230 [+] on Build GRCh38
          GenBankmRNA DA932306.1 - 3 444 Chr. 1 - 28506044 - 28509327 [+] on Build GRCh38
          GenBankmRNA BG028447.1 - 3 428 Chr. 1 - 28506057 - 28509522 [+] on Build GRCh38
          GenBankmRNA CB128113.1 - 3 447 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA BG291633.1 - 3 426 Chr. 1 - 28506067 - 28509489 [+] on Build GRCh38
          GenBankmRNA AI004003.1 - 1 276 Chr. 1 - 28508923 - 28509326 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 428 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA BI833122.1 - 3 427 Chr. 1 - 28506059 - 28509542 [+] on Build GRCh38
          GenBankmRNA DA224254.1 - 3 446 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA926278.1 - 3 445 Chr. 1 - 28506044 - 28509334 [+] on Build GRCh38
          GenBankmRNA DA056610.1 - 3 503 Chr. 1 - 28505984 - 28509252 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 432 Chr. - - [-] on Build
          GenBankmRNA BU198833.1 - 3 448 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA H54484.1 - 1 70 Chr. 1 - 28509130 - 28509544 [+] on Build GRCh38
          GenBankmRNA BF036396.1 - 2 410 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA DA652687.1 - 3 446 Chr. 1 - 28506044 - 28509333 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 428 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0