Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          SNHG3
          Gene Name:
          small nucleolar RNA host gene 3
          Gene Aliases:
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          UniGene ID:
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258486

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 616
          NR_036473.1
          4 724
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 516
          AJ006835.1 - LIT1315
          U17HG
          3 516
          AK092726.1 -
          2 2249
          AK311577.1 -
          1 268
          BC006790.1 -
          3 498
          BC012122.1 -
          3 504
          BC017334.2 -
          3 491
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM479709.1 - 3 501 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA DA820174.1 - 3 513 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 469 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA DA925005.1 - 3 515 Chr. 1 - 28506044 - 28509319 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 493 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 492 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 503 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA DA936462.1 - 3 515 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 493 Chr. - - [-] on Build
          GenBankmRNA DB112428.1 - 3 516 Chr. 1 - 28506044 - 28509353 [+] on Build GRCh38
          GenBankmRNA DA998284.1 - 3 524 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 485 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 487 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA DA952260.1 - 3 516 Chr. 1 - 28506044 - 28509313 [+] on Build GRCh38
          GenBankmRNA CN366542.1 - 4 452 Chr. 1 - 28506055 - 28509581 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 519 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 394 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 450 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA DA565956.1 - 3 515 Chr. 1 - 28506044 - 28509324 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 499 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 515 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA DB113121.1 - 3 516 Chr. 1 - 28506044 - 28509329 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 516 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA DA928567.1 - 3 513 Chr. 1 - 28506047 - 28509337 [+] on Build GRCh38
          GenBankmRNA DB262011.1 - 3 516 Chr. 1 - 28506044 - 28509330 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 501 Chr. - - [-] on Build
          GenBankmRNA DB271395.1 - 3 516 Chr. 1 - 28506044 - 28509295 [+] on Build GRCh38
          GenBankmRNA DA943290.1 - 3 517 Chr. 1 - 28506043 - 28509321 [+] on Build GRCh38
          GenBankmRNA DW417474.1 - 161 Chr. - - [-] on Build
          GenBankmRNA CB985655.1 - 3 519 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 511 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 570 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA DA969742.1 - 3 520 Chr. 1 - 28506045 - 28509315 [+] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 516 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA DA968918.1 - 3 515 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BF982554.1 - 2 461 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 506 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA BG023899.1 - 484 Chr. - - [-] on Build
          GenBankmRNA BM790281.1 - 3 503 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 534 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA DA902617.1 - 3 515 Chr. 1 - 28506045 - 28509297 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 515 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 513 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA CD652956.1 - 1 143 Chr. 1 - 28509129 - 28509633 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 484 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA EB387088.1 - 1 177 Chr. 1 - 28509094 - 28509634 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 515 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 559 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 515 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA DA932306.1 - 3 515 Chr. 1 - 28506044 - 28509327 [+] on Build GRCh38
          GenBankmRNA DB024065.1 - 1 268 Chr. 1 - 28509002 - 28509575 [+] on Build GRCh38
          GenBankmRNA DA918003.1 - 3 516 Chr. 1 - 28506044 - 28509337 [+] on Build GRCh38
          GenBankmRNA DA817644.1 - 3 516 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA847021.1 - 3 516 Chr. 1 - 28506044 - 28509325 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 515 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 488 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 499 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA AI004003.1 - 1 347 Chr. 1 - 28508923 - 28509326 [+] on Build GRCh38
          GenBankmRNA DA926278.1 - 3 516 Chr. 1 - 28506044 - 28509334 [+] on Build GRCh38
          GenBankmRNA DA224254.1 - 3 517 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 498 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA961704.1 - 3 516 Chr. 1 - 28506044 - 28509303 [+] on Build GRCh38
          GenBankmRNA BU198833.1 - 3 519 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 503 Chr. - - [-] on Build
          GenBankmRNA H54484.1 - 1 141 Chr. 1 - 28509130 - 28509544 [+] on Build GRCh38
          GenBankmRNA DA922155.1 - 3 516 Chr. 1 - 28506044 - 28509320 [+] on Build GRCh38
          GenBankmRNA BF036396.1 - 2 481 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA DA652687.1 - 3 517 Chr. 1 - 28506044 - 28509333 [+] on Build GRCh38
          GenBankmRNA DB157161.1 - 3 517 Chr. 1 - 28506043 - 28509332 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 499 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38
          GenBankmRNA DA644956.1 - 3 516 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0