Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          SNHG3 RCC1
          Gene Name:
          small nucleolar RNA host gene 3
          regulator of chromosome condensation 1
          Gene Aliases:
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          CHC1, RCC1-I, SNHG3-RCC1
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          Chromosome Location:
          Chr.1: 28505943 - 28539196 on Build GRCh38
          UniGene ID:
          Hs.469723
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n258490

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 426
          NR_036473.1
          4 534
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 326
          AJ006835.1 - LIT1315
          U17HG
          3 326
          AK092726.1 -
          2 2059
          AK311577.1 -
          1 79
          BC006790.1 -
          3 308
          BC012122.1 -
          3 314
          BC017334.2 -
          3 301
          DA044266.1 -
          3 385
          DA135739.1 -
          4 497
          Gene Symbol: RCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BF667678.1 - 3 425
          CB985055.1 - 3 392
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA820174.1 - 3 323 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY075800.1 - 3 326 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA AA352342.1 - 2 277 Chr. 1 - 28508136 - 28509153 [+] on Build GRCh38
          GenBankmRNA HY180673.1 - 3 324 Chr. 1 - 28506045 - 28509192 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 303 Chr. - - [-] on Build
          GenBankmRNA DA998284.1 - 3 334 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA CN366542.1 - 4 263 Chr. 1 - 28506055 - 28509581 [+] on Build GRCh38
          GenBankmRNA DA955189.1 - 3 328 Chr. 1 - 28506043 - 28509267 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 260 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA DB113121.1 - 3 326 Chr. 1 - 28506044 - 28509329 [+] on Build GRCh38
          GenBankmRNA BF941702.1 - 3 318 Chr. 1 - 28506050 - 28509152 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 326 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA HY182836.1 - 3 324 Chr. 1 - 28506045 - 28509242 [+] on Build GRCh38
          GenBankmRNA AA352343.1 - 2 277 Chr. 1 - 28508136 - 28509163 [+] on Build GRCh38
          GenBankmRNA DA928567.1 - 3 323 Chr. 1 - 28506047 - 28509337 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 312 Chr. - - [-] on Build
          GenBankmRNA HY112100.1 - 3 400 Chr. 1 - 28505969 - 28509173 [+] on Build GRCh38
          GenBankmRNA CB985655.1 - 3 329 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 322 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA DA969742.1 - 3 330 Chr. 1 - 28506045 - 28509315 [+] on Build GRCh38
          GenBankmRNA BM802102.1 - 313 Chr. - - [-] on Build
          GenBankmRNA DA968918.1 - 3 325 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM790281.1 - 3 313 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA AV763089.1 - 2 319 Chr. 1 - 28508127 - 28509528 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 345 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 325 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA DA089660.1 - 3 386 Chr. 1 - 28505984 - 28509274 [+] on Build GRCh38
          GenBankmRNA HY176129.1 - 3 324 Chr. 1 - 28506045 - 28509283 [+] on Build GRCh38
          GenBankmRNA DA704998.1 - 3 325 Chr. 1 - 28506044 - 28509252 [+] on Build GRCh38
          GenBankmRNA AV689452.1 - 3 360 Chr. 1 - 28506009 - 28509366 [+] on Build GRCh38
          GenBankmRNA CN366541.1 - 2 282 Chr. 1 - 28508130 - 28509185 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 325 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 325 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA BE867363.1 - 2 299 Chr. 1 - 28508131 - 28509511 [+] on Build GRCh38
          GenBankmRNA DB024065.1 - 1 79 Chr. 1 - 28509002 - 28509575 [+] on Build GRCh38
          GenBankmRNA DA918003.1 - 3 326 Chr. 1 - 28506044 - 28509337 [+] on Build GRCh38
          GenBankmRNA DA817644.1 - 3 326 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA847021.1 - 3 326 Chr. 1 - 28506044 - 28509325 [+] on Build GRCh38
          GenBankmRNA HY177570.1 - 3 324 Chr. 1 - 28506045 - 28509225 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 298 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 325 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 308 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA961704.1 - 3 326 Chr. 1 - 28506044 - 28509303 [+] on Build GRCh38
          GenBankmRNA HY179875.1 - 3 323 Chr. 1 - 28506045 - 28509263 [+] on Build GRCh38
          GenBankmRNA DA922155.1 - 3 326 Chr. 1 - 28506044 - 28509320 [+] on Build GRCh38
          GenBankmRNA AV764560.2 - 3 319 Chr. 1 - 28506051 - 28509412 [+] on Build GRCh38
          GenBankmRNA DB157161.1 - 3 327 Chr. 1 - 28506043 - 28509332 [+] on Build GRCh38
          GenBankmRNA DA644956.1 - 3 326 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA BM479709.1 - 3 311 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 280 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA HY173512.1 - 3 325 Chr. 1 - 28506045 - 28509232 [+] on Build GRCh38
          GenBankmRNA DA925005.1 - 3 325 Chr. 1 - 28506044 - 28509319 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 305 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 313 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 302 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA DA936462.1 - 3 325 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DB112428.1 - 3 326 Chr. 1 - 28506044 - 28509353 [+] on Build GRCh38
          GenBankmRNA HY113224.1 - 3 382 Chr. 1 - 28505989 - 28509206 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 295 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 297 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA DA952260.1 - 3 326 Chr. 1 - 28506044 - 28509313 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 204 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 329 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY153255.1 - 3 324 Chr. 1 - 28506045 - 28509198 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 310 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA DA565956.1 - 3 325 Chr. 1 - 28506044 - 28509324 [+] on Build GRCh38
          GenBankmRNA HY084156.1 - 3 324 Chr. 1 - 28506045 - 28509217 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 325 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA BG474710.1 - 3 316 Chr. 1 - 28506055 - 28509618 [+] on Build GRCh38
          GenBankmRNA CB157773.1 - 3 314 Chr. 1 - 28506055 - 28509221 [+] on Build GRCh38
          GenBankmRNA DB262011.1 - 3 326 Chr. 1 - 28506044 - 28509330 [+] on Build GRCh38
          GenBankmRNA DB271395.1 - 3 326 Chr. 1 - 28506044 - 28509295 [+] on Build GRCh38
          GenBankmRNA DA943290.1 - 3 327 Chr. 1 - 28506043 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY089919.1 - 3 324 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 380 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA HY179907.1 - 3 324 Chr. 1 - 28506045 - 28509261 [+] on Build GRCh38
          GenBankmRNA DA510712.1 - 3 326 Chr. 1 - 28506044 - 28509216 [+] on Build GRCh38
          GenBankmRNA AK022892.1 - 1 1325 Chr. 22 - 49888443 - 49891419 [-] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 326 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA BF982554.1 - 2 273 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 316 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA DA787816.1 - 3 325 Chr. 1 - 28506044 - 28509270 [+] on Build GRCh38
          GenBankmRNA BG023899.1 - 296 Chr. - - [-] on Build
          GenBankmRNA DA902617.1 - 3 325 Chr. 1 - 28506045 - 28509297 [+] on Build GRCh38
          GenBankmRNA HY176989.1 - 3 323 Chr. 1 - 28506045 - 28509229 [+] on Build GRCh38
          GenBankmRNA HY175139.1 - 3 324 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 323 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 294 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 369 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA CN366539.1 - 3 301 Chr. 1 - 28506068 - 28509242 [+] on Build GRCh38
          GenBankmRNA HY182945.1 - 3 324 Chr. 1 - 28506045 - 28509230 [+] on Build GRCh38
          GenBankmRNA DA932306.1 - 3 325 Chr. 1 - 28506044 - 28509327 [+] on Build GRCh38
          GenBankmRNA BG028447.1 - 3 311 Chr. 1 - 28506057 - 28509522 [+] on Build GRCh38
          GenBankmRNA CB128113.1 - 3 328 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA BG291633.1 - 3 307 Chr. 1 - 28506067 - 28509489 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 309 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA AI004003.1 - 1 158 Chr. 1 - 28508923 - 28509326 [+] on Build GRCh38
          GenBankmRNA BI833122.1 - 3 308 Chr. 1 - 28506059 - 28509542 [+] on Build GRCh38
          GenBankmRNA DA224254.1 - 3 327 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA926278.1 - 3 326 Chr. 1 - 28506044 - 28509334 [+] on Build GRCh38
          GenBankmRNA DA056610.1 - 3 385 Chr. 1 - 28505984 - 28509252 [+] on Build GRCh38
          GenBankmRNA BU198833.1 - 3 329 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 313 Chr. - - [-] on Build
          GenBankmRNA BF036396.1 - 2 291 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA DA652687.1 - 3 327 Chr. 1 - 28506044 - 28509333 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 309 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0