Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          SNHG3 RCC1
          Gene Name:
          small nucleolar RNA host gene 3
          regulator of chromosome condensation 1
          Gene Aliases:
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          CHC1, RCC1-I, SNHG3-RCC1
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          Chromosome Location:
          Chr.1: 28505943 - 28539196 on Build GRCh38
          UniGene ID:
          Hs.469723
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n258491

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 488
          NR_036473.1
          4 596
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 388
          AJ006835.1 - LIT1315
          U17HG
          3 388
          AK092726.1 -
          2 2121
          AK311577.1 -
          1 141
          BC006790.1 -
          3 370
          BC012122.1 -
          3 376
          BC017334.2 -
          3 363
          DA044266.1 -
          3 447
          Gene Symbol: RCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA CB985055.1 - 3 455
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA820174.1 - 3 385 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY075800.1 - 3 388 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA AA352342.1 - 2 339 Chr. 1 - 28508136 - 28509153 [+] on Build GRCh38
          GenBankmRNA HY180673.1 - 3 386 Chr. 1 - 28506045 - 28509192 [+] on Build GRCh38
          GenBankmRNA HY147758.1 - 3 393 Chr. 1 - 28506045 - 28509206 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 365 Chr. - - [-] on Build
          GenBankmRNA DA998284.1 - 3 396 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA CN366542.1 - 4 325 Chr. 1 - 28506055 - 28509581 [+] on Build GRCh38
          GenBankmRNA DA955189.1 - 3 390 Chr. 1 - 28506043 - 28509267 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 322 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA EH333588.1 - 1 54 Chr. 1 - 28509088 - 28509163 [+] on Build GRCh38
          GenBankmRNA DB113121.1 - 3 388 Chr. 1 - 28506044 - 28509329 [+] on Build GRCh38
          GenBankmRNA BF941702.1 - 3 380 Chr. 1 - 28506050 - 28509152 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 388 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA HY182836.1 - 3 386 Chr. 1 - 28506045 - 28509242 [+] on Build GRCh38
          GenBankmRNA AA352343.1 - 2 341 Chr. 1 - 28508136 - 28509163 [+] on Build GRCh38
          GenBankmRNA DA928567.1 - 3 385 Chr. 1 - 28506047 - 28509337 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 374 Chr. - - [-] on Build
          GenBankmRNA HY112100.1 - 3 462 Chr. 1 - 28505969 - 28509173 [+] on Build GRCh38
          GenBankmRNA DW417474.1 - 33 Chr. - - [-] on Build
          GenBankmRNA CB985655.1 - 3 391 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 384 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA DA969742.1 - 3 392 Chr. 1 - 28506045 - 28509315 [+] on Build GRCh38
          GenBankmRNA DA968918.1 - 3 387 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM802102.1 - 375 Chr. - - [-] on Build
          GenBankmRNA BM790281.1 - 3 375 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 407 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA AV763089.1 - 2 381 Chr. 1 - 28508127 - 28509528 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 387 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA DA089660.1 - 3 448 Chr. 1 - 28505984 - 28509274 [+] on Build GRCh38
          GenBankmRNA HY176129.1 - 3 386 Chr. 1 - 28506045 - 28509283 [+] on Build GRCh38
          GenBankmRNA EB387088.1 - 1 49 Chr. 1 - 28509094 - 28509634 [+] on Build GRCh38
          GenBankmRNA DA704998.1 - 3 387 Chr. 1 - 28506044 - 28509252 [+] on Build GRCh38
          GenBankmRNA CN366541.1 - 2 344 Chr. 1 - 28508130 - 28509185 [+] on Build GRCh38
          GenBankmRNA AV689452.1 - 3 418 Chr. 1 - 28506009 - 28509366 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 387 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 387 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA BE867363.1 - 2 360 Chr. 1 - 28508131 - 28509511 [+] on Build GRCh38
          GenBankmRNA DB024065.1 - 1 141 Chr. 1 - 28509002 - 28509575 [+] on Build GRCh38
          GenBankmRNA DA918003.1 - 3 388 Chr. 1 - 28506044 - 28509337 [+] on Build GRCh38
          GenBankmRNA BG115368.1 - 2 383 Chr. 1 - 28508127 - 28509208 [+] on Build GRCh38
          GenBankmRNA DA817644.1 - 3 388 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA847021.1 - 3 388 Chr. 1 - 28506044 - 28509325 [+] on Build GRCh38
          GenBankmRNA HY177570.1 - 3 386 Chr. 1 - 28506045 - 28509225 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 360 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 387 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 370 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA961704.1 - 3 388 Chr. 1 - 28506044 - 28509303 [+] on Build GRCh38
          GenBankmRNA HY179875.1 - 3 385 Chr. 1 - 28506045 - 28509263 [+] on Build GRCh38
          GenBankmRNA DA922155.1 - 3 388 Chr. 1 - 28506044 - 28509320 [+] on Build GRCh38
          GenBankmRNA AV764560.2 - 3 381 Chr. 1 - 28506051 - 28509412 [+] on Build GRCh38
          GenBankmRNA DB157161.1 - 3 389 Chr. 1 - 28506043 - 28509332 [+] on Build GRCh38
          GenBankmRNA DA644956.1 - 3 388 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA BM479709.1 - 3 373 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 342 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA HY173512.1 - 3 387 Chr. 1 - 28506045 - 28509232 [+] on Build GRCh38
          GenBankmRNA DA925005.1 - 3 387 Chr. 1 - 28506044 - 28509319 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 367 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 364 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 375 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA DA936462.1 - 3 387 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DB112428.1 - 3 388 Chr. 1 - 28506044 - 28509353 [+] on Build GRCh38
          GenBankmRNA HY113224.1 - 3 444 Chr. 1 - 28505989 - 28509206 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 357 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 359 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA DA952260.1 - 3 388 Chr. 1 - 28506044 - 28509313 [+] on Build GRCh38
          GenBankmRNA BE841288.1 - 1 74 Chr. 1 - 28509097 - 28509254 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 391 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 266 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA HY153255.1 - 3 386 Chr. 1 - 28506045 - 28509198 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 372 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA DA565956.1 - 3 387 Chr. 1 - 28506044 - 28509324 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 387 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA HY084156.1 - 3 386 Chr. 1 - 28506045 - 28509217 [+] on Build GRCh38
          GenBankmRNA BG474710.1 - 3 378 Chr. 1 - 28506055 - 28509618 [+] on Build GRCh38
          GenBankmRNA CB157773.1 - 3 376 Chr. 1 - 28506055 - 28509221 [+] on Build GRCh38
          GenBankmRNA DB262011.1 - 3 388 Chr. 1 - 28506044 - 28509330 [+] on Build GRCh38
          GenBankmRNA DA943290.1 - 3 389 Chr. 1 - 28506043 - 28509321 [+] on Build GRCh38
          GenBankmRNA DB271395.1 - 3 388 Chr. 1 - 28506044 - 28509295 [+] on Build GRCh38
          GenBankmRNA HY089919.1 - 3 386 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 442 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA HY179907.1 - 3 386 Chr. 1 - 28506045 - 28509261 [+] on Build GRCh38
          GenBankmRNA DA510712.1 - 3 388 Chr. 1 - 28506044 - 28509216 [+] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 388 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA BF982554.1 - 2 335 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA DA787816.1 - 3 387 Chr. 1 - 28506044 - 28509270 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 378 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA BG023899.1 - 358 Chr. - - [-] on Build
          GenBankmRNA DA902617.1 - 3 387 Chr. 1 - 28506045 - 28509297 [+] on Build GRCh38
          GenBankmRNA HY176989.1 - 3 385 Chr. 1 - 28506045 - 28509229 [+] on Build GRCh38
          GenBankmRNA HY175139.1 - 3 386 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 385 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA CD652956.1 - 1 15 Chr. 1 - 28509129 - 28509633 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 356 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 431 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA CN366539.1 - 3 363 Chr. 1 - 28506068 - 28509242 [+] on Build GRCh38
          GenBankmRNA HY182945.1 - 3 386 Chr. 1 - 28506045 - 28509230 [+] on Build GRCh38
          GenBankmRNA DA932306.1 - 3 387 Chr. 1 - 28506044 - 28509327 [+] on Build GRCh38
          GenBankmRNA FN072314.1 - 1 68 Chr. 1 - 28509071 - 28509176 [+] on Build GRCh38
          GenBankmRNA BG028447.1 - 3 373 Chr. 1 - 28506057 - 28509522 [+] on Build GRCh38
          GenBankmRNA CB128113.1 - 3 390 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA BG291633.1 - 3 369 Chr. 1 - 28506067 - 28509489 [+] on Build GRCh38
          GenBankmRNA AI004003.1 - 1 220 Chr. 1 - 28508923 - 28509326 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 371 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA BI833122.1 - 3 370 Chr. 1 - 28506059 - 28509542 [+] on Build GRCh38
          GenBankmRNA DA926278.1 - 3 388 Chr. 1 - 28506044 - 28509334 [+] on Build GRCh38
          GenBankmRNA DA224254.1 - 3 389 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA056610.1 - 3 447 Chr. 1 - 28505984 - 28509252 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 375 Chr. - - [-] on Build
          GenBankmRNA BU198833.1 - 3 391 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA H54484.1 - 1 13 Chr. 1 - 28509130 - 28509544 [+] on Build GRCh38
          GenBankmRNA BF036396.1 - 2 353 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA DA652687.1 - 3 389 Chr. 1 - 28506044 - 28509333 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 371 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0