Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          SNHG3 RCC1
          Gene Name:
          small nucleolar RNA host gene 3
          regulator of chromosome condensation 1
          Gene Aliases:
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          CHC1, RCC1-I, SNHG3-RCC1
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          Chromosome Location:
          Chr.1: 28505943 - 28539196 on Build GRCh38
          UniGene ID:
          Hs.469723
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n258492

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 390
          NR_036473.1
          4 498
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 290
          AJ006835.1 - LIT1315
          U17HG
          3 290
          AK092726.1 -
          2 2023
          AK311577.1 -
          1 43
          BC006790.1 -
          3 272
          BC012122.1 -
          3 278
          BC017334.2 -
          3 265
          DA044266.1 -
          3 349
          DA135739.1 -
          4 461
          Gene Symbol: RCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA CB985055.1 - 3 356
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA820174.1 - 3 287 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY075800.1 - 3 290 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA AA352342.1 - 2 241 Chr. 1 - 28508136 - 28509153 [+] on Build GRCh38
          GenBankmRNA HY180673.1 - 3 288 Chr. 1 - 28506045 - 28509192 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 267 Chr. - - [-] on Build
          GenBankmRNA DA998284.1 - 3 298 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA CN366542.1 - 4 227 Chr. 1 - 28506055 - 28509581 [+] on Build GRCh38
          GenBankmRNA DA955189.1 - 3 292 Chr. 1 - 28506043 - 28509267 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 224 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA DB113121.1 - 3 290 Chr. 1 - 28506044 - 28509329 [+] on Build GRCh38
          GenBankmRNA BF941702.1 - 3 282 Chr. 1 - 28506050 - 28509152 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 290 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA HY182836.1 - 3 288 Chr. 1 - 28506045 - 28509242 [+] on Build GRCh38
          GenBankmRNA AA352343.1 - 2 241 Chr. 1 - 28508136 - 28509163 [+] on Build GRCh38
          GenBankmRNA DA928567.1 - 3 287 Chr. 1 - 28506047 - 28509337 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 276 Chr. - - [-] on Build
          GenBankmRNA HY112100.1 - 3 364 Chr. 1 - 28505969 - 28509173 [+] on Build GRCh38
          GenBankmRNA CB985655.1 - 3 293 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 286 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA DA969742.1 - 3 294 Chr. 1 - 28506045 - 28509315 [+] on Build GRCh38
          GenBankmRNA DA968918.1 - 3 289 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM802102.1 - 277 Chr. - - [-] on Build
          GenBankmRNA BM790281.1 - 3 277 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 309 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA AV763089.1 - 2 283 Chr. 1 - 28508127 - 28509528 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 289 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA DA089660.1 - 3 350 Chr. 1 - 28505984 - 28509274 [+] on Build GRCh38
          GenBankmRNA HY176129.1 - 3 288 Chr. 1 - 28506045 - 28509283 [+] on Build GRCh38
          GenBankmRNA DA704998.1 - 3 289 Chr. 1 - 28506044 - 28509252 [+] on Build GRCh38
          GenBankmRNA CN366541.1 - 2 246 Chr. 1 - 28508130 - 28509185 [+] on Build GRCh38
          GenBankmRNA AV689452.1 - 3 324 Chr. 1 - 28506009 - 28509366 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 289 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 289 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA BE867363.1 - 2 263 Chr. 1 - 28508131 - 28509511 [+] on Build GRCh38
          GenBankmRNA DB024065.1 - 1 43 Chr. 1 - 28509002 - 28509575 [+] on Build GRCh38
          GenBankmRNA DA918003.1 - 3 290 Chr. 1 - 28506044 - 28509337 [+] on Build GRCh38
          GenBankmRNA DA817644.1 - 3 290 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA847021.1 - 3 290 Chr. 1 - 28506044 - 28509325 [+] on Build GRCh38
          GenBankmRNA HY177570.1 - 3 288 Chr. 1 - 28506045 - 28509225 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 262 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 289 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 272 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA961704.1 - 3 290 Chr. 1 - 28506044 - 28509303 [+] on Build GRCh38
          GenBankmRNA HY179875.1 - 3 287 Chr. 1 - 28506045 - 28509263 [+] on Build GRCh38
          GenBankmRNA DA922155.1 - 3 290 Chr. 1 - 28506044 - 28509320 [+] on Build GRCh38
          GenBankmRNA AV764560.2 - 3 283 Chr. 1 - 28506051 - 28509412 [+] on Build GRCh38
          GenBankmRNA DB157161.1 - 3 291 Chr. 1 - 28506043 - 28509332 [+] on Build GRCh38
          GenBankmRNA DA644956.1 - 3 290 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA BM479709.1 - 3 275 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 244 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA HY173512.1 - 3 289 Chr. 1 - 28506045 - 28509232 [+] on Build GRCh38
          GenBankmRNA DA925005.1 - 3 289 Chr. 1 - 28506044 - 28509319 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 269 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 277 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 266 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA DA936462.1 - 3 289 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DB112428.1 - 3 290 Chr. 1 - 28506044 - 28509353 [+] on Build GRCh38
          GenBankmRNA HY113224.1 - 3 346 Chr. 1 - 28505989 - 28509206 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 259 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 261 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA DA952260.1 - 3 290 Chr. 1 - 28506044 - 28509313 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 293 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 168 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA HY153255.1 - 3 288 Chr. 1 - 28506045 - 28509198 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 274 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA DA565956.1 - 3 289 Chr. 1 - 28506044 - 28509324 [+] on Build GRCh38
          GenBankmRNA HY084156.1 - 3 288 Chr. 1 - 28506045 - 28509217 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 289 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA BG474710.1 - 3 280 Chr. 1 - 28506055 - 28509618 [+] on Build GRCh38
          GenBankmRNA CB157773.1 - 3 278 Chr. 1 - 28506055 - 28509221 [+] on Build GRCh38
          GenBankmRNA DB262011.1 - 3 290 Chr. 1 - 28506044 - 28509330 [+] on Build GRCh38
          GenBankmRNA DB271395.1 - 3 290 Chr. 1 - 28506044 - 28509295 [+] on Build GRCh38
          GenBankmRNA DA943290.1 - 3 291 Chr. 1 - 28506043 - 28509321 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 344 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA HY179907.1 - 3 288 Chr. 1 - 28506045 - 28509261 [+] on Build GRCh38
          GenBankmRNA DA510712.1 - 3 290 Chr. 1 - 28506044 - 28509216 [+] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 290 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA BF982554.1 - 2 237 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 280 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA DA787816.1 - 3 289 Chr. 1 - 28506044 - 28509270 [+] on Build GRCh38
          GenBankmRNA BG023899.1 - 259 Chr. - - [-] on Build
          GenBankmRNA DA902617.1 - 3 289 Chr. 1 - 28506045 - 28509297 [+] on Build GRCh38
          GenBankmRNA HY176989.1 - 3 287 Chr. 1 - 28506045 - 28509229 [+] on Build GRCh38
          GenBankmRNA HY175139.1 - 3 288 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 287 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 258 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 333 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA CN366539.1 - 3 265 Chr. 1 - 28506068 - 28509242 [+] on Build GRCh38
          GenBankmRNA HY182945.1 - 3 288 Chr. 1 - 28506045 - 28509230 [+] on Build GRCh38
          GenBankmRNA DA932306.1 - 3 289 Chr. 1 - 28506044 - 28509327 [+] on Build GRCh38
          GenBankmRNA BG028447.1 - 3 275 Chr. 1 - 28506057 - 28509522 [+] on Build GRCh38
          GenBankmRNA CB128113.1 - 3 292 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA AI004003.1 - 1 122 Chr. 1 - 28508923 - 28509326 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 273 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA BI833122.1 - 3 272 Chr. 1 - 28506059 - 28509542 [+] on Build GRCh38
          GenBankmRNA DA926278.1 - 3 290 Chr. 1 - 28506044 - 28509334 [+] on Build GRCh38
          GenBankmRNA DA224254.1 - 3 291 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA056610.1 - 3 349 Chr. 1 - 28505984 - 28509252 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 277 Chr. - - [-] on Build
          GenBankmRNA BU198833.1 - 3 293 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA BF036396.1 - 2 255 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA DA652687.1 - 3 291 Chr. 1 - 28506044 - 28509333 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 273 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dsplp:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0