Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG3 SIRNA Assays ›
          Gene Symbol:
          SNHG3 RCC1
          Gene Name:
          small nucleolar RNA host gene 3
          regulator of chromosome condensation 1
          Gene Aliases:
          NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
          CHC1, RCC1-I, SNHG3-RCC1
          Chromosome Location:
          Chr.1: 28505943 - 28510892 on Build GRCh38
          Chromosome Location:
          Chr.1: 28505943 - 28539196 on Build GRCh38
          UniGene ID:
          Hs.469723
          Hs.469723
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID n258493

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002909.2
          3 341
          NR_036473.1
          4 449
          GenBank mRNA AJ006834.1 - LIT1316
          U17HG
          3 241
          AJ006835.1 - LIT1315
          U17HG
          3 241
          AK092726.1 -
          2 1974
          BC006790.1 -
          3 223
          BC012122.1 -
          3 229
          BC017334.2 -
          3 216
          DA044266.1 -
          3 300
          DA135739.1 -
          4 412
          Gene Symbol: RCC1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA CB985055.1 - 3 307
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA820174.1 - 3 238 Chr. 1 - 28506046 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY075800.1 - 3 241 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA AA352342.1 - 2 192 Chr. 1 - 28508136 - 28509153 [+] on Build GRCh38
          GenBankmRNA HY180673.1 - 3 239 Chr. 1 - 28506045 - 28509192 [+] on Build GRCh38
          GenBankmRNA BM450342.1 - 218 Chr. - - [-] on Build
          GenBankmRNA DA998284.1 - 3 249 Chr. 1 - 28506045 - 28509318 [+] on Build GRCh38
          GenBankmRNA DA955189.1 - 3 243 Chr. 1 - 28506043 - 28509267 [+] on Build GRCh38
          GenBankmRNA DA933716.1 - 1 175 Chr. 1 - 28508824 - 28509359 [+] on Build GRCh38
          GenBankmRNA DB113121.1 - 3 241 Chr. 1 - 28506044 - 28509329 [+] on Build GRCh38
          GenBankmRNA BF941702.1 - 3 233 Chr. 1 - 28506050 - 28509152 [+] on Build GRCh38
          GenBankmRNA CN366538.1 - 3 241 Chr. 1 - 28506055 - 28509488 [+] on Build GRCh38
          GenBankmRNA HY182836.1 - 3 239 Chr. 1 - 28506045 - 28509242 [+] on Build GRCh38
          GenBankmRNA AA352343.1 - 2 192 Chr. 1 - 28508136 - 28509163 [+] on Build GRCh38
          GenBankmRNA DA928567.1 - 3 238 Chr. 1 - 28506047 - 28509337 [+] on Build GRCh38
          GenBankmRNA BF795420.1 - 227 Chr. - - [-] on Build
          GenBankmRNA HY112100.1 - 3 315 Chr. 1 - 28505969 - 28509173 [+] on Build GRCh38
          GenBankmRNA CB985655.1 - 3 244 Chr. 1 - 28506043 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY059835.1 - 3 237 Chr. 1 - 28506045 - 28509325 [+] on Build GRCh38
          GenBankmRNA DA969742.1 - 3 245 Chr. 1 - 28506045 - 28509315 [+] on Build GRCh38
          GenBankmRNA BM802102.1 - 228 Chr. - - [-] on Build
          GenBankmRNA DA968918.1 - 3 240 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA BM790281.1 - 3 228 Chr. 1 - 28506056 - 28509323 [+] on Build GRCh38
          GenBankmRNA AV763089.1 - 2 234 Chr. 1 - 28508127 - 28509528 [+] on Build GRCh38
          GenBankmRNA CD048541.1 - 3 260 Chr. 1 - 28506045 - 28509614 [+] on Build GRCh38
          GenBankmRNA DA478694.1 - 3 240 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA DA089660.1 - 3 301 Chr. 1 - 28505984 - 28509274 [+] on Build GRCh38
          GenBankmRNA HY176129.1 - 3 239 Chr. 1 - 28506045 - 28509283 [+] on Build GRCh38
          GenBankmRNA DA704998.1 - 3 240 Chr. 1 - 28506044 - 28509252 [+] on Build GRCh38
          GenBankmRNA CN366541.1 - 2 197 Chr. 1 - 28508130 - 28509185 [+] on Build GRCh38
          GenBankmRNA AV689452.1 - 3 275 Chr. 1 - 28506009 - 28509366 [+] on Build GRCh38
          GenBankmRNA DA728986.1 - 3 240 Chr. 1 - 28506044 - 28509467 [+] on Build GRCh38
          GenBankmRNA BQ224438.1 - 3 240 Chr. 1 - 28506055 - 28509617 [+] on Build GRCh38
          GenBankmRNA DA918003.1 - 3 241 Chr. 1 - 28506044 - 28509337 [+] on Build GRCh38
          GenBankmRNA DA817644.1 - 3 241 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA847021.1 - 3 241 Chr. 1 - 28506044 - 28509325 [+] on Build GRCh38
          GenBankmRNA HY177570.1 - 3 239 Chr. 1 - 28506045 - 28509225 [+] on Build GRCh38
          GenBankmRNA BU187480.1 - 2 213 Chr. 1 - 28508127 - 28509634 [+] on Build GRCh38
          GenBankmRNA BM771983.1 - 3 240 Chr. 1 - 28506044 - 28509374 [+] on Build GRCh38
          GenBankmRNA BI772556.1 - 2 223 Chr. 1 - 28508116 - 28509484 [+] on Build GRCh38
          GenBankmRNA DA961704.1 - 3 241 Chr. 1 - 28506044 - 28509303 [+] on Build GRCh38
          GenBankmRNA HY179875.1 - 3 238 Chr. 1 - 28506045 - 28509263 [+] on Build GRCh38
          GenBankmRNA DA922155.1 - 3 241 Chr. 1 - 28506044 - 28509320 [+] on Build GRCh38
          GenBankmRNA AV764560.2 - 3 234 Chr. 1 - 28506051 - 28509412 [+] on Build GRCh38
          GenBankmRNA DB157161.1 - 3 242 Chr. 1 - 28506043 - 28509332 [+] on Build GRCh38
          GenBankmRNA DA644956.1 - 3 241 Chr. 1 - 28506044 - 28509328 [+] on Build GRCh38
          GenBankmRNA BM479709.1 - 3 226 Chr. 1 - 28506059 - 28509634 [+] on Build GRCh38
          GenBankmRNA CN366536.1 - 2 195 Chr. 1 - 28508133 - 28509423 [+] on Build GRCh38
          GenBankmRNA HY173512.1 - 3 240 Chr. 1 - 28506045 - 28509232 [+] on Build GRCh38
          GenBankmRNA DA925005.1 - 3 240 Chr. 1 - 28506044 - 28509319 [+] on Build GRCh38
          GenBankmRNA BI833284.1 - 3 220 Chr. 1 - 28506060 - 28509531 [+] on Build GRCh38
          GenBankmRNA BU183513.1 - 3 228 Chr. 1 - 28506056 - 28509509 [+] on Build GRCh38
          GenBankmRNA BM788818.1 - 2 217 Chr. 1 - 28508127 - 28509339 [+] on Build GRCh38
          GenBankmRNA DA936462.1 - 3 240 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DB112428.1 - 3 241 Chr. 1 - 28506044 - 28509353 [+] on Build GRCh38
          GenBankmRNA HY113224.1 - 3 297 Chr. 1 - 28505989 - 28509206 [+] on Build GRCh38
          GenBankmRNA AL523116.3 - 2 212 Chr. 1 - 28508127 - 28509940 [+] on Build GRCh38
          GenBankmRNA CN366544.1 - 2 210 Chr. 1 - 28508127 - 28509283 [+] on Build GRCh38
          GenBankmRNA DA952260.1 - 3 241 Chr. 1 - 28506044 - 28509313 [+] on Build GRCh38
          GenBankmRNA BM834932.1 - 1 119 Chr. 1 - 28508876 - 28509340 [+] on Build GRCh38
          GenBankmRNA BU943197.1 - 3 244 Chr. 1 - 28506044 - 28509509 [+] on Build GRCh38
          GenBankmRNA HY153255.1 - 3 239 Chr. 1 - 28506045 - 28509198 [+] on Build GRCh38
          GenBankmRNA CN366540.1 - 2 225 Chr. 1 - 28508127 - 28509421 [+] on Build GRCh38
          GenBankmRNA DA565956.1 - 3 240 Chr. 1 - 28506044 - 28509324 [+] on Build GRCh38
          GenBankmRNA HY084156.1 - 3 239 Chr. 1 - 28506045 - 28509217 [+] on Build GRCh38
          GenBankmRNA DB128729.1 - 3 240 Chr. 1 - 28506044 - 28509326 [+] on Build GRCh38
          GenBankmRNA BG474710.1 - 3 231 Chr. 1 - 28506055 - 28509618 [+] on Build GRCh38
          GenBankmRNA CB157773.1 - 3 229 Chr. 1 - 28506055 - 28509221 [+] on Build GRCh38
          GenBankmRNA DB262011.1 - 3 241 Chr. 1 - 28506044 - 28509330 [+] on Build GRCh38
          GenBankmRNA DB271395.1 - 3 241 Chr. 1 - 28506044 - 28509295 [+] on Build GRCh38
          GenBankmRNA DA943290.1 - 3 242 Chr. 1 - 28506043 - 28509321 [+] on Build GRCh38
          GenBankmRNA HY089919.1 - 3 239 Chr. 1 - 28506045 - 28509246 [+] on Build GRCh38
          GenBankmRNA CB963044.1 - 3 295 Chr. 1 - 28505990 - 28509462 [+] on Build GRCh38
          GenBankmRNA HY179907.1 - 3 239 Chr. 1 - 28506045 - 28509261 [+] on Build GRCh38
          GenBankmRNA DA510712.1 - 3 241 Chr. 1 - 28506044 - 28509216 [+] on Build GRCh38
          GenBankmRNA BM759606.1 - 3 241 Chr. 1 - 28506043 - 28509424 [+] on Build GRCh38
          GenBankmRNA BF982554.1 - 2 188 Chr. 1 - 28508137 - 28509570 [+] on Build GRCh38
          GenBankmRNA DA787816.1 - 3 240 Chr. 1 - 28506044 - 28509270 [+] on Build GRCh38
          GenBankmRNA CN366535.1 - 3 231 Chr. 1 - 28506053 - 28509489 [+] on Build GRCh38
          GenBankmRNA DA902617.1 - 3 240 Chr. 1 - 28506045 - 28509297 [+] on Build GRCh38
          GenBankmRNA HY176989.1 - 3 238 Chr. 1 - 28506045 - 28509229 [+] on Build GRCh38
          GenBankmRNA HY175139.1 - 3 239 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA CN366543.1 - 3 238 Chr. 1 - 28506055 - 28509366 [+] on Build GRCh38
          GenBankmRNA BY796950.2 - 2 209 Chr. 1 - 28508127 - 28509414 [+] on Build GRCh38
          GenBankmRNA DA381375.1 - 1 284 Chr. 1 - 28508764 - 28509340 [+] on Build GRCh38
          GenBankmRNA CN366539.1 - 3 216 Chr. 1 - 28506068 - 28509242 [+] on Build GRCh38
          GenBankmRNA HY182945.1 - 3 239 Chr. 1 - 28506045 - 28509230 [+] on Build GRCh38
          GenBankmRNA DA932306.1 - 3 240 Chr. 1 - 28506044 - 28509327 [+] on Build GRCh38
          GenBankmRNA BG028447.1 - 3 226 Chr. 1 - 28506057 - 28509522 [+] on Build GRCh38
          GenBankmRNA CB128113.1 - 3 243 Chr. 1 - 28506045 - 28509219 [+] on Build GRCh38
          GenBankmRNA AI004003.1 - 1 73 Chr. 1 - 28508923 - 28509326 [+] on Build GRCh38
          GenBankmRNA CB163464.1 - 3 224 Chr. 1 - 28506060 - 28509328 [+] on Build GRCh38
          GenBankmRNA BI833122.1 - 3 223 Chr. 1 - 28506059 - 28509542 [+] on Build GRCh38
          GenBankmRNA DA926278.1 - 3 241 Chr. 1 - 28506044 - 28509334 [+] on Build GRCh38
          GenBankmRNA DA224254.1 - 3 242 Chr. 1 - 28506044 - 28509331 [+] on Build GRCh38
          GenBankmRNA DA056610.1 - 3 300 Chr. 1 - 28505984 - 28509252 [+] on Build GRCh38
          GenBankmRNA BU198833.1 - 3 244 Chr. 1 - 28506044 - 28509422 [+] on Build GRCh38
          GenBankmRNA BM802666.1 - 228 Chr. - - [-] on Build
          GenBankmRNA BF036396.1 - 2 206 Chr. 1 - 28508127 - 28509376 [+] on Build GRCh38
          GenBankmRNA DA652687.1 - 3 242 Chr. 1 - 28506044 - 28509333 [+] on Build GRCh38
          GenBankmRNA BQ218931.1 - 3 224 Chr. 1 - 28506060 - 28509544 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-r46r8:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0