Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other API5 SIRNA Assays ›
          Gene Symbol:
          API5
          Gene Name:
          apoptosis inhibitor 5
          Gene Aliases:
          AAC-11, AAC11
          Chromosome Location:
          Chr.11: 43311955 - 43344530 on Build GRCh38
          UniGene ID:
          Hs.435771
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258666

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: API5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024625.1 2 286
          XM_017018464.1 XP_016873953.1 2 271
          XM_017018465.1 XP_016873954.1 2 271
          GenBank mRNA AK304157.1 - 2 252
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC416340.1 - 2 255 Chr. 11 - 43311989 - 43321434 [+] on Build GRCh38
          GenBankmRNA DC417519.1 - 2 245 Chr. 11 - 43311996 - 43321477 [+] on Build GRCh38
          GenBankmRNA DC309550.1 - 2 252 Chr. 11 - 43311989 - 43321465 [+] on Build GRCh38
          GenBankmRNA CF138429.1 - 2 232 Chr. 11 - 43312014 - 43320836 [+] on Build GRCh38
          GenBankmRNA DC416862.1 - 2 245 Chr. 11 - 43311998 - 43321479 [+] on Build GRCh38
          GenBankmRNA DC335814.1 - 2 252 Chr. 11 - 43311989 - 43321464 [+] on Build GRCh38
          GenBankmRNA DC404208.1 - 2 209 Chr. 11 - 43312032 - 43322039 [+] on Build GRCh38
          GenBankmRNA DC333712.1 - 2 254 Chr. 11 - 43311989 - 43321464 [+] on Build GRCh38
          GenBankmRNA DC415796.1 - 2 253 Chr. 11 - 43311989 - 43318803 [+] on Build GRCh38
          GenBankmRNA DC423851.1 - 2 263 Chr. 11 - 43311978 - 43321439 [+] on Build GRCh38
          GenBankmRNA DC325081.1 - 2 206 Chr. 11 - 43312035 - 43322031 [+] on Build GRCh38
          GenBankmRNA CD243789.1 - 2 244 Chr. 11 - 43311997 - 43322137 [+] on Build GRCh38
          GenBankmRNA DC414185.1 - 2 252 Chr. 11 - 43311989 - 43321459 [+] on Build GRCh38
          GenBankmRNA DC306792.1 - 2 252 Chr. 11 - 43311989 - 43321433 [+] on Build GRCh38
          GenBankmRNA DC423666.1 - 2 245 Chr. 11 - 43311996 - 43321467 [+] on Build GRCh38
          GenBankmRNA DC401496.1 - 2 245 Chr. 11 - 43311996 - 43321444 [+] on Build GRCh38
          GenBankmRNA DC416512.1 - 2 252 Chr. 11 - 43311989 - 43321462 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          negative regulation of apoptotic process
          negative regulation of fibroblast apoptotic process
          protein binding
          fibroblast growth factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-z8nhc:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0