Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other STK16 SIRNA Assays ›
          Gene Symbol:
          STK16
          Gene Name:
          serine/threonine kinase 16
          Gene Aliases:
          KRCT, MPSK, PKL12, TSF1
          Chromosome Location:
          Chr.2: 219245469 - 219250337 on Build GRCh38
          UniGene ID:
          Hs.153003
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258747

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: STK16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026909.1 1 136
          XM_017005160.1 XP_016860649.1 1 105
          GenBank mRNA AK096954.1 - 1 108
          AK302182.1 - 1 105
          AK310793.1 - 1 136
          AL552406.3 - 1 113
          CB111662.1 - 1 145
          CN268850.1 - 1 85
          DB190729.1 - 1 136
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI834709.1 - 1 80 Chr. 2 - 219245535 - 219246876 [+] on Build GRCh38
          GenBankmRNA BP329123.1 - 1 81 Chr. 2 - 219245534 - 219246784 [+] on Build GRCh38
          GenBankmRNA BI755751.1 - 1 71 Chr. 2 - 219245544 - 219247488 [+] on Build GRCh38
          GenBankmRNA BI828291.1 - 1 58 Chr. 2 - 219245558 - 219247631 [+] on Build GRCh38
          GenBankmRNA DA751927.1 - 1 110 Chr. 2 - 219245505 - 219246734 [+] on Build GRCh38
          GenBankmRNA DA552837.1 - 1 110 Chr. 2 - 219245505 - 219246164 [+] on Build GRCh38
          GenBankmRNA DB004053.1 - 1 110 Chr. 2 - 219245505 - 219246139 [+] on Build GRCh38
          GenBankmRNA AL711197.1 - 1 95 Chr. 2 - 219245521 - 219246103 [+] on Build GRCh38
          GenBankmRNA HY037477.1 - 1 82 Chr. 2 - 219245534 - 219246085 [+] on Build GRCh38
          GenBankmRNA BI916892.1 - 1 97 Chr. 2 - 219245518 - 219247151 [+] on Build GRCh38
          GenBankmRNA DB048250.1 - 1 105 Chr. 2 - 219245510 - 219246741 [+] on Build GRCh38
          GenBankmRNA BI761409.1 - 1 90 Chr. 2 - 219245525 - 219247230 [+] on Build GRCh38
          GenBankmRNA BI834773.1 - 1 81 Chr. 2 - 219245535 - 219247432 [+] on Build GRCh38
          GenBankmRNA BX495596.1 - 1 90 Chr. 2 - 219245525 - 219246157 [+] on Build GRCh38
          GenBankmRNA BI830567.1 - 1 80 Chr. 2 - 219245535 - 219247494 [+] on Build GRCh38
          GenBankmRNA DA696612.1 - 1 120 Chr. 2 - 219245495 - 219246047 [+] on Build GRCh38
          GenBankmRNA DA612050.1 - 1 119 Chr. 2 - 219245496 - 219246714 [+] on Build GRCh38
          GenBankmRNA BX443461.2 - 1 98 Chr. 2 - 219245517 - 219247722 [+] on Build GRCh38
          GenBankmRNA BX324712.2 - 1 85 Chr. 2 - 219245536 - 219247235 [+] on Build GRCh38
          GenBankmRNA BM543451.1 - 125 Chr. - - [-] on Build
          GenBankmRNA BX332722.2 - 1 87 Chr. 2 - 219245529 - 219247535 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription from RNA polymerase II promoter
          peptidyl-tyrosine phosphorylation
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          cellular response to transforming growth factor beta stimulus
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          protein serine/threonine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein binding
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0