Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC4P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002773.1 | 1 | 1729 | |||
GenBank mRNA | AF047485.1 | - | 1 | 1729 | ||
AF047486.1 | - | 2 | 518 |