Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPP1R3E SIRNA Assays ›
          Gene Symbol:
          PPP1R3E
          Gene Name:
          protein phosphatase 1 regulatory subunit 3E
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 23295921 - 23302848 on Build GRCh38
          UniGene ID:
          Hs.601513
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n259040

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPP1R3E
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276318.1 NP_001263247.1 4 1470
          GenBank mRNA AK024489.1 - 4 1470
          AK091943.1 - 3 362
          AK296016.1 - 2 238
          BC039400.1 - 4 534
          BX161476.1 - 2 933
          BX248758.1 - 2 947
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BQ186915.1 - 4 331 Chr. 14 - 23298585 - 23301475 [-] on Build GRCh38
          GenBankmRNA BX408826.2 - 2 229 Chr. 14 - 23298301 - 23300219 [-] on Build GRCh38
          GenBankmRNA DB167626.1 - 3 238 Chr. 14 - 23298606 - 23302883 [-] on Build GRCh38
          GenBankmRNA DA450655.1 - 2 290 Chr. 14 - 23298660 - 23301183 [-] on Build GRCh38
          GenBankmRNA DA361739.1 - 2 238 Chr. 14 - 23298627 - 23301525 [-] on Build GRCh38
          GenBankmRNA D63235.1 - 2 303 Chr. 14 - 23298745 - 23301489 [-] on Build GRCh38
          GenBankmRNA BX429404.2 - 1 114 Chr. 14 - 23298477 - 23299542 [-] on Build GRCh38
          GenBankmRNA AA167648.1 - 3 342 Chr. 14 - 23298720 - 23301499 [-] on Build GRCh38
          GenBankmRNA DA182949.1 - 2 345 Chr. 14 - 23298728 - 23301237 [-] on Build GRCh38
          GenBankmRNA BX347011.1 - 2 396 Chr. 14 - 23298477 - 23301036 [-] on Build GRCh38
          GenBankmRNA DA796287.1 - 4 358 Chr. 14 - 23298758 - 23301502 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphatase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen metabolic process
          regulation of glycogen biosynthetic process
          regulation of glycogen catabolic process
          protein dephosphorylation
          phosphoprotein phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0