Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBIL5P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024120.2 | 1 | 2278 | |||
GenBank mRNA | AF229804.1 | - | 1 | 1951 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX092190.1 | - | 2 | 562 | Chr. 17 - 752667 - 754755 [+] on Build GRCh38 |