Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMCX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028407.2 | 16 | 3683 | |||
NR_045861.1 | 12 | 3340 | ||||
NR_045862.1 | 12 | 2703 | ||||
NR_045864.1 | 12 | 3382 | ||||
GenBank mRNA | AK125041.1 | - | 12 | 2501 | ||
AK126987.1 | - | 12 | 3203 | |||
BC032236.1 | - | 12 | 3119 | |||
CR933662.1 | - | 16 | 3658 |