Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSBP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024448.2 | 10 | 2775 | |||
GenBank mRNA | AK025313.1 | - | 1 | 271 | ||
AK056875.1 | - | 10 | 2705 | |||
AK091837.1 | - | 9 | 2513 | |||
AK125808.1 | - | 3 | 3175 | |||
BC063045.1 | - | 8 | 1018 | |||
BX538181.1 | - | 10 | 2669 | |||
CR933599.1 | - | 2 | 3047 | |||
S82637.1 | - | 2 | 267 | |||
X52204.1 | - | 2 | 269 |