Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PART1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024617.1 | 1 | 297 | |||
NR_028508.1 | 1 | 78 | ||||
NR_028509.1 | 1 | 78 | ||||
GenBank mRNA | AF163475.1 | - | 1 | 78 | ||
AK312071.1 | - | 1 | 300 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA296238.1 | - | 1 | 33 | Chr. 5 - 60487977 - 60529508 [+] on Build GRCh38 |