Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RHNO1 SIRNA Assays ›
          Gene Symbol:
          RHNO1
          Gene Name:
          RAD9-HUS1-RAD1 interacting nuclear orphan 1
          Gene Aliases:
          C12orf32, HKMT1188, RHINO
          Chromosome Location:
          Chr.12: 2876258 - 2889525 on Build GRCh38
          UniGene ID:
          Hs.198853
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260682

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RHNO1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252499.2 NP_001239428.1 3 452
          NM_001252500.2 NP_001239429.1 3 410
          NM_001257097.1 NP_001244026.1 3 573
          NM_001257098.1 NP_001244027.1 3 472
          NR_027365.2 2 200
          NR_046432.1 4 586
          NR_046433.1 2 374
          GenBank mRNA AB073599.1 - 3 428
          AK021945.1 - 2 176
          AK304613.1 - 3 386
          AK315364.1 - 3 436
          BC000079.1 - 1 26
          BC005106.1 - 3 418
          BM847461.1 - 2 375
          DA679606.1 - 3 472
          DA749974.1 - 3 549
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX478096.1 - 3 435 Chr. 12 - 2877217 - 2888194 [+] on Build GRCh38
          GenBankmRNA HY002442.1 - 3 433 Chr. 12 - 2877220 - 2888059 [+] on Build GRCh38
          GenBankmRNA DC399643.1 - 3 410 Chr. 12 - 2877199 - 2888159 [+] on Build GRCh38
          GenBankmRNA BX951872.1 - 3 443 Chr. 12 - 2877209 - 2888047 [+] on Build GRCh38
          GenBankmRNA CB960794.1 - 3 475 Chr. 12 - 2877203 - 2888266 [+] on Build GRCh38
          GenBankmRNA HY136095.1 - 3 391 Chr. 12 - 2877220 - 2888059 [+] on Build GRCh38
          GenBankmRNA DA524829.1 - 3 434 Chr. 12 - 2877217 - 2888129 [+] on Build GRCh38
          GenBankmRNA DA022625.1 - 3 434 Chr. 12 - 2877217 - 2888157 [+] on Build GRCh38
          GenBankmRNA DA923751.1 - 3 452 Chr. 12 - 2877199 - 2888133 [+] on Build GRCh38
          GenBankmRNA DA914577.1 - 3 452 Chr. 12 - 2877199 - 2888128 [+] on Build GRCh38
          GenBankmRNA BI517428.1 - 3 443 Chr. 12 - 2877208 - 2888488 [+] on Build GRCh38
          GenBankmRNA CD704714.1 - 3 465 Chr. 12 - 2877220 - 2888145 [+] on Build GRCh38
          GenBankmRNA BQ224346.1 - 438 Chr. - - [-] on Build
          GenBankmRNA DA426269.1 - 3 433 Chr. 12 - 2877218 - 2888158 [+] on Build GRCh38
          GenBankmRNA BX952162.1 - 3 432 Chr. 12 - 2877220 - 2888241 [+] on Build GRCh38
          GenBankmRNA HY006186.1 - 3 432 Chr. 12 - 2877220 - 2888059 [+] on Build GRCh38
          GenBankmRNA HY127825.1 - 3 390 Chr. 12 - 2877220 - 2888114 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          DNA damage checkpoint
          recombinational repair
          DNA replication
          cellular response to UV
          positive regulation of G0 to G1 transition
          cellular response to ionizing radiation
          regulation of signal transduction by p53 class mediator
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0