Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNRD1ASP SIRNA Assays ›
          Gene Symbol:
          ZNRD1ASP
          Gene Name:
          zinc ribbon domain containing 1 antisense, pseudogene
          Gene Aliases:
          C6orf12, HTEX4, NCRNA00171, TCTEX4, ZNRD1-AS, ZNRD1-AS1, ZNRD1AS, ZNRD1AS1
          Chromosome Location:
          Chr.6: 30001011 - 30061184 on Build GRCh38
          UniGene ID:
          Hs.653168
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260735

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNRD1ASP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026751.1 4 475
          GenBank mRNA AF032110.1 - 4 1191
          AF238323.1 - 3 1013
          BC067890.1 - 4 475
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BI462223.1 - 4 481 Chr. 6 - 30002658 - 30061184 [-] on Build GRCh38
          GenBankmRNA DB079640.1 - 3 297 Chr. 6 - 30021723 - 30061144 [-] on Build GRCh38
          GenBankmRNA DA107135.1 - 4 448 Chr. 6 - 30035038 - 30061157 [-] on Build GRCh38
          GenBankmRNA BQ423269.1 - 3 469 Chr. 6 - 30021686 - 30061095 [-] on Build GRCh38
          GenBankmRNA DA517191.1 - 4 407 Chr. 6 - 30035024 - 30061116 [-] on Build GRCh38
          GenBankmRNA DB096670.1 - 4 435 Chr. 6 - 30035023 - 30061144 [-] on Build GRCh38
          GenBankmRNA DA099095.1 - 4 469 Chr. 6 - 30035068 - 30061178 [-] on Build GRCh38
          GenBankmRNA DB030030.1 - 3 159 Chr. 6 - 29834970 - 30060784 [-] on Build GRCh38
          GenBankmRNA DA015305.1 - 4 479 Chr. 6 - 30035041 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA329344.1 - 3 214 Chr. 6 - 30009812 - 30060840 [-] on Build GRCh38
          GenBankmRNA DB453565.2 - 4 406 Chr. 6 - 30023736 - 30061141 [-] on Build GRCh38
          GenBankmRNA HY018590.1 - 4 472 Chr. 6 - 30035928 - 30061206 [-] on Build GRCh38
          GenBankmRNA BQ434771.1 - 3 544 Chr. 6 - 30021686 - 30061169 [-] on Build GRCh38
          GenBankmRNA AL705833.1 - 3 494 Chr. 6 - 30034935 - 30058747 [-] on Build GRCh38
          GenBankmRNA DB061771.1 - 4 477 Chr. 6 - 30035039 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA734967.1 - 448 Chr. - - [-] on Build
          GenBankmRNA DA507524.1 - 3 391 Chr. 6 - 30034963 - 30061017 [-] on Build GRCh38
          GenBankmRNA BG421163.1 - 420 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xq6xf:80/100.66.76.183:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0