Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZNRD1ASP SIRNA Assays ›
          Gene Symbol:
          ZNRD1ASP
          Gene Name:
          zinc ribbon domain containing 1 antisense, pseudogene
          Gene Aliases:
          C6orf12, HTEX4, NCRNA00171, TCTEX4, ZNRD1-AS, ZNRD1-AS1, ZNRD1AS, ZNRD1AS1
          Chromosome Location:
          Chr.6: 30001011 - 30061184 on Build GRCh38
          UniGene ID:
          Hs.653168
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260737

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZNRD1ASP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026751.1 2 348
          GenBank mRNA AF032109.2 - 1 226
          AF032110.1 - 1 436
          AF238323.1 - 1 886
          BC067890.1 - 2 348
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA699002.1 - 1 419 Chr. 6 - 30009812 - 30061171 [-] on Build GRCh38
          GenBankmRNA AL547982.3 - 1 390 Chr. 6 - 30002364 - 30061143 [-] on Build GRCh38
          GenBankmRNA DB278897.1 - 2 349 Chr. 6 - 30021657 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA056456.1 - 2 332 Chr. 6 - 30007358 - 30061184 [-] on Build GRCh38
          GenBankmRNA BQ423269.1 - 1 342 Chr. 6 - 30021686 - 30061095 [-] on Build GRCh38
          GenBankmRNA DB096670.1 - 2 308 Chr. 6 - 30035023 - 30061144 [-] on Build GRCh38
          GenBankmRNA DA082317.1 - 2 348 Chr. 6 - 30016047 - 30061184 [-] on Build GRCh38
          GenBankmRNA DB030030.1 - 1 31 Chr. 6 - 29834970 - 30060784 [-] on Build GRCh38
          GenBankmRNA DA255293.1 - 1 404 Chr. 6 - 30035915 - 30061157 [-] on Build GRCh38
          GenBankmRNA BE744018.1 - 1 175 Chr. 6 - 30002183 - 30060928 [-] on Build GRCh38
          GenBankmRNA DA329344.1 - 1 87 Chr. 6 - 30009812 - 30060840 [-] on Build GRCh38
          GenBankmRNA DB453565.2 - 2 279 Chr. 6 - 30023736 - 30061141 [-] on Build GRCh38
          GenBankmRNA HY018590.1 - 2 345 Chr. 6 - 30035928 - 30061206 [-] on Build GRCh38
          GenBankmRNA DA965516.1 - 2 348 Chr. 6 - 30007530 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA064369.1 - 1 404 Chr. 6 - 30035034 - 30061157 [-] on Build GRCh38
          GenBankmRNA DB065301.1 - 1 431 Chr. 6 - 30058136 - 30061184 [-] on Build GRCh38
          GenBankmRNA DB061771.1 - 2 350 Chr. 6 - 30035039 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA507524.1 - 1 264 Chr. 6 - 30034963 - 30061017 [-] on Build GRCh38
          GenBankmRNA DA527722.1 - 1 278 Chr. 6 - 29927506 - 30061031 [-] on Build GRCh38
          GenBankmRNA DA107135.1 - 2 321 Chr. 6 - 30035038 - 30061157 [-] on Build GRCh38
          GenBankmRNA DB096928.1 - 2 352 Chr. 6 - 30057587 - 30061188 [-] on Build GRCh38
          GenBankmRNA DA517191.1 - 2 280 Chr. 6 - 30035024 - 30061116 [-] on Build GRCh38
          GenBankmRNA DA411941.1 - 1 432 Chr. 6 - 30035937 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA099095.1 - 2 342 Chr. 6 - 30035068 - 30061178 [-] on Build GRCh38
          GenBankmRNA DA492271.1 - 1 428 Chr. 6 - 30002676 - 30061183 [-] on Build GRCh38
          GenBankmRNA DA015305.1 - 2 352 Chr. 6 - 30035041 - 30061184 [-] on Build GRCh38
          GenBankmRNA DA502551.1 - 2 280 Chr. 6 - 30007564 - 30061116 [-] on Build GRCh38
          GenBankmRNA DA091689.1 - 1 411 Chr. 6 - 30021689 - 30061163 [-] on Build GRCh38
          GenBankmRNA DA514793.1 - 1 281 Chr. 6 - 30007336 - 30061034 [-] on Build GRCh38
          GenBankmRNA DB064495.1 - 2 348 Chr. 6 - 30009784 - 30061184 [-] on Build GRCh38
          GenBankmRNA BQ434771.1 - 1 417 Chr. 6 - 30021686 - 30061169 [-] on Build GRCh38
          GenBankmRNA DB222151.1 - 2 321 Chr. 6 - 30009754 - 30061157 [-] on Build GRCh38
          GenBankmRNA DA734967.1 - 321 Chr. - - [-] on Build
          GenBankmRNA BG421163.1 - 293 Chr. - - [-] on Build

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0