Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAXIP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024476.1 | 4 | 953 | |||
NR_024477.1 | 4 | 953 | ||||
GenBank mRNA | AK056609.1 | - | 4 | 943 | ||
AK311721.1 | - | 3 | 876 | |||
BC033053.1 | - | 4 | 979 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CA389913.1 | - | 4 | 593 | Chr. 7 - 154928655 - 154946448 [+] on Build GRCh38 |