Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001448.1 | 10 | 1003 | |||
GenBank mRNA | AJ132821.1 | - | 9 | 937 | ||
AJ132822.1 | - | 9 | 819 | |||
AJ132823.1 | - | 8 | 753 | |||
BC047448.1 | - | 10 | 1137 | |||
BC067864.1 | - | 8 | 796 |