Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016430.3 | NP_057514.2 | 3, 4 | 426 | ||
XM_017028365.1 | XP_016883854.1 | 3, 4 | 368 | |||
GenBank mRNA | AB037164.1 | - | 2, 3 | 277 | ||
AF216305.1 | - | 3, 4 | 342 |