Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC01278 SIRNA Assays ›
          Gene Symbol:
          LINC01278
          Gene Name:
          long intergenic non-protein coding RNA 1278
          Gene Aliases:
          -
          Chromosome Location:
          Chr.X: 63426559 - 63560993 on Build GRCh38
          UniGene ID:
          Hs.31532
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261216

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC01278
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015353.1 4 546
          GenBank mRNA AK001756.1 - 4 546
          AK130294.1 - 1 2749
          AL080202.1 - 3 429
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA510286.1 - 3 440 Chr. X - 63428752 - 63561060 [-] on Build GRCh38
          GenBankmRNA DB131712.1 - 3 423 Chr. X - 63428870 - 63561040 [-] on Build GRCh38
          GenBankmRNA DA178606.1 - 3 440 Chr. X - 63428870 - 63561060 [-] on Build GRCh38
          GenBankmRNA DA918322.1 - 3 403 Chr. X - 63428733 - 63561023 [-] on Build GRCh38
          GenBankmRNA DA699810.1 - 3 425 Chr. X - 63428801 - 63561045 [-] on Build GRCh38
          GenBankmRNA DA746517.1 - 3 403 Chr. X - 63428736 - 63561023 [-] on Build GRCh38
          GenBankmRNA DA763164.1 - 3 413 Chr. X - 63428739 - 63561033 [-] on Build GRCh38
          GenBankmRNA DA630098.1 - 3 420 Chr. X - 63428773 - 63561040 [-] on Build GRCh38
          GenBankmRNA DB254947.1 - 3 403 Chr. X - 63428731 - 63561023 [-] on Build GRCh38
          GenBankmRNA AU135375.1 - 3 244 Chr. X - 63428527 - 63516942 [-] on Build GRCh38
          GenBankmRNA DB120212.1 - 3 432 Chr. X - 63428778 - 63561052 [-] on Build GRCh38
          GenBankmRNA DA768320.1 - 3 450 Chr. X - 63428829 - 63561070 [-] on Build GRCh38
          GenBankmRNA DA708197.1 - 3 451 Chr. X - 63428863 - 63561071 [-] on Build GRCh38
          GenBankmRNA DB250520.1 - 3 403 Chr. X - 63428710 - 63561023 [-] on Build GRCh38
          GenBankmRNA DN997080.1 - 3 391 Chr. X - 63428490 - 63561005 [-] on Build GRCh38
          GenBankmRNA CA394355.1 - 3 340 Chr. X - 63428527 - 63517038 [-] on Build GRCh38
          GenBankmRNA DA843572.1 - 3 316 Chr. X - 63428636 - 63434393 [-] on Build GRCh38
          GenBankmRNA DA739870.1 - 3 417 Chr. X - 63428784 - 63561037 [-] on Build GRCh38
          GenBankmRNA DA586399.1 - 3 451 Chr. X - 63428751 - 63561070 [-] on Build GRCh38
          GenBankmRNA DA923582.1 - 1 300 Chr. X - 63428612 - 63429198 [-] on Build GRCh38
          GenBankmRNA DA451054.1 - 3 455 Chr. X - 63428793 - 63561075 [-] on Build GRCh38
          GenBankmRNA DB207443.1 - 3 403 Chr. X - 63428749 - 63561023 [-] on Build GRCh38
          GenBankmRNA BU182259.1 - 2 214 Chr. X - 63428228 - 63432471 [-] on Build GRCh38
          GenBankmRNA DA775097.1 - 2 129 Chr. X - 63428475 - 63432409 [-] on Build GRCh38
          GenBankmRNA DA370924.1 - 3 431 Chr. X - 63428778 - 63561050 [-] on Build GRCh38
          GenBankmRNA DA848272.1 - 3 316 Chr. X - 63428623 - 63434392 [-] on Build GRCh38
          GenBankmRNA DA474747.1 - 3 425 Chr. X - 63428761 - 63561045 [-] on Build GRCh38
          GenBankmRNA AU133450.1 - 4 546 Chr. X - 63428593 - 63560993 [-] on Build GRCh38
          GenBankmRNA DA846337.1 - 3 304 Chr. X - 63428620 - 63434381 [-] on Build GRCh38
          GenBankmRNA DA742553.1 - 4 485 Chr. X - 63428820 - 63560932 [-] on Build GRCh38
          GenBankmRNA DB263371.1 - 3 451 Chr. X - 63428772 - 63561071 [-] on Build GRCh38
          GenBankmRNA DA518360.1 - 1 470 Chr. X - 63428803 - 63429368 [-] on Build GRCh38
          GenBankmRNA CN360625.1 - 4 518 Chr. X - 63428876 - 63560965 [-] on Build GRCh38
          GenBankmRNA DA148199.1 - 3 404 Chr. X - 63428735 - 63561023 [-] on Build GRCh38
          GenBankmRNA DA867657.1 - 1 529 Chr. X - 63428883 - 63429426 [-] on Build GRCh38
          GenBankmRNA DB158545.1 - 3 478 Chr. X - 63428784 - 63561098 [-] on Build GRCh38
          GenBankmRNA CN360628.1 - 4 405 Chr. X - 63428554 - 63560853 [-] on Build GRCh38
          GenBankmRNA DA534104.1 - 3 416 Chr. X - 63428760 - 63561036 [-] on Build GRCh38
          GenBankmRNA BQ432093.1 - 3 481 Chr. X - 63428483 - 63561057 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0