Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WAC SIRNA Assays ›
          Gene Symbol:
          WAC
          Gene Name:
          WW domain containing adaptor with coiled-coil
          Gene Aliases:
          BM-016, DESSH, PRO1741, Wwp4
          Chromosome Location:
          Chr.10: 28532426 - 28623112 on Build GRCh38
          UniGene ID:
          Hs.743224
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261282

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WAC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_100264.2 NP_567822.1 1 369
          XM_011519491.2 XP_011517793.1 1 531
          XM_017016315.1 XP_016871804.1 1 189
          XM_017016316.1 XP_016871805.1 1 531
          XM_017016317.1 XP_016871806.1 1 189
          XM_017016318.1 XP_016871807.1 1 189
          GenBank mRNA AK055852.1 - 1 186
          AK225846.1 - 1 182
          BI463901.1 - 1 469
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC424507.1 - 1 182 Chr. 10 - 28532775 - 28590832 [+] on Build GRCh38
          GenBankmRNA DC358674.1 - 1 185 Chr. 10 - 28532775 - 28583464 [+] on Build GRCh38
          GenBankmRNA DA786316.1 - 1 190 Chr. 10 - 28532773 - 28583497 [+] on Build GRCh38
          GenBankmRNA DA791734.1 - 1 182 Chr. 10 - 28532775 - 28583491 [+] on Build GRCh38
          GenBankmRNA DC389074.1 - 1 206 Chr. 10 - 28532752 - 28583461 [+] on Build GRCh38
          GenBankmRNA AW956436.1 - 1 116 Chr. 10 - 28532841 - 28535601 [+] on Build GRCh38
          GenBankmRNA DC365418.1 - 1 186 Chr. 10 - 28532773 - 28583482 [+] on Build GRCh38
          GenBankmRNA DC425084.1 - 1 185 Chr. 10 - 28532772 - 28583479 [+] on Build GRCh38
          GenBankmRNA DA045431.1 - 1 182 Chr. 10 - 28532775 - 28583505 [+] on Build GRCh38
          GenBankmRNA CB070240.1 - 1 73 Chr. 10 - 28532884 - 28583420 [+] on Build GRCh38
          GenBankmRNA BM759552.1 - 1 178 Chr. 10 - 28532779 - 28589780 [+] on Build GRCh38
          GenBankmRNA DA200076.1 - 1 99 Chr. 10 - 28532858 - 28589772 [+] on Build GRCh38
          GenBankmRNA DC376383.1 - 1 189 Chr. 10 - 28532768 - 28583497 [+] on Build GRCh38
          GenBankmRNA DC416902.1 - 1 182 Chr. 10 - 28532775 - 28583469 [+] on Build GRCh38
          GenBankmRNA DA735921.1 - 1 171 Chr. 10 - 28532786 - 28590809 [+] on Build GRCh38
          GenBankmRNA DA921274.1 - 1 178 Chr. 10 - 28532779 - 28583489 [+] on Build GRCh38
          GenBankmRNA DA256377.1 - 1 185 Chr. 10 - 28532773 - 28583494 [+] on Build GRCh38
          GenBankmRNA DR155734.1 - 1 178 Chr. 10 - 28532791 - 28590832 [+] on Build GRCh38
          GenBankmRNA DA223735.1 - 1 178 Chr. 10 - 28532779 - 28535758 [+] on Build GRCh38
          GenBankmRNA DA850839.1 - 1 178 Chr. 10 - 28532779 - 28583476 [+] on Build GRCh38
          GenBankmRNA DA084741.1 - 1 182 Chr. 10 - 28532776 - 28535960 [+] on Build GRCh38
          GenBankmRNA DA113057.1 - 1 178 Chr. 10 - 28532779 - 28589767 [+] on Build GRCh38
          GenBankmRNA DA219361.1 - 1 183 Chr. 10 - 28532776 - 28535755 [+] on Build GRCh38
          GenBankmRNA DB135981.1 - 1 185 Chr. 10 - 28532772 - 28583478 [+] on Build GRCh38
          GenBankmRNA DC366045.1 - 1 184 Chr. 10 - 28532776 - 28583458 [+] on Build GRCh38
          GenBankmRNA DB098040.1 - 1 183 Chr. 10 - 28532776 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA590921.1 - 1 182 Chr. 10 - 28532775 - 28583499 [+] on Build GRCh38
          GenBankmRNA DA575859.1 - 1 151 Chr. 10 - 28532806 - 28583505 [+] on Build GRCh38
          GenBankmRNA DC298540.1 - 1 183 Chr. 10 - 28532776 - 28583503 [+] on Build GRCh38
          GenBankmRNA DA069792.1 - 1 178 Chr. 10 - 28532779 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA216611.1 - 1 182 Chr. 10 - 28532775 - 28583484 [+] on Build GRCh38
          GenBankmRNA DA920864.1 - 1 187 Chr. 10 - 28532771 - 28583487 [+] on Build GRCh38
          GenBankmRNA DB248503.1 - 1 183 Chr. 10 - 28532774 - 28589772 [+] on Build GRCh38
          GenBankmRNA DA528199.1 - 1 185 Chr. 10 - 28532773 - 28589758 [+] on Build GRCh38
          GenBankmRNA DC361394.1 - 1 181 Chr. 10 - 28532776 - 28583459 [+] on Build GRCh38
          GenBankmRNA DA818719.1 - 1 183 Chr. 10 - 28532774 - 28583490 [+] on Build GRCh38
          GenBankmRNA DC328307.1 - 1 203 Chr. 10 - 28532754 - 28583492 [+] on Build GRCh38
          GenBankmRNA DA016395.1 - 1 178 Chr. 10 - 28532779 - 28583490 [+] on Build GRCh38
          GenBankmRNA DB281041.1 - 1 183 Chr. 10 - 28532777 - 28583482 [+] on Build GRCh38
          GenBankmRNA DA817988.1 - 1 185 Chr. 10 - 28532773 - 28583491 [+] on Build GRCh38
          GenBankmRNA DA198111.1 - 1 187 Chr. 10 - 28532773 - 28583495 [+] on Build GRCh38
          GenBankmRNA DC358172.1 - 1 183 Chr. 10 - 28532776 - 28590783 [+] on Build GRCh38
          GenBankmRNA DA931274.1 - 1 182 Chr. 10 - 28532775 - 28590794 [+] on Build GRCh38
          GenBankmRNA DA682593.1 - 1 187 Chr. 10 - 28532772 - 28535758 [+] on Build GRCh38
          GenBankmRNA DC299187.1 - 1 182 Chr. 10 - 28532775 - 28583491 [+] on Build GRCh38
          GenBankmRNA DC318888.1 - 1 178 Chr. 10 - 28532779 - 28583472 [+] on Build GRCh38
          GenBankmRNA DA247908.1 - 1 182 Chr. 10 - 28532775 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA293258.1 - 1 180 Chr. 10 - 28532779 - 28583505 [+] on Build GRCh38
          GenBankmRNA DC319805.1 - 1 183 Chr. 10 - 28532774 - 28583489 [+] on Build GRCh38
          GenBankmRNA DC375626.1 - 1 201 Chr. 10 - 28532756 - 28583501 [+] on Build GRCh38
          GenBankmRNA BE886618.1 - 1 114 Chr. 10 - 28532843 - 28595781 [+] on Build GRCh38
          GenBankmRNA DA241504.1 - 1 182 Chr. 10 - 28532775 - 28583496 [+] on Build GRCh38
          GenBankmRNA DA713840.1 - 1 173 Chr. 10 - 28532784 - 28583504 [+] on Build GRCh38
          GenBankmRNA DA995456.1 - 1 185 Chr. 10 - 28532772 - 28535761 [+] on Build GRCh38
          GenBankmRNA DC346912.1 - 1 184 Chr. 10 - 28532773 - 28583477 [+] on Build GRCh38
          GenBankmRNA DA634496.1 - 1 182 Chr. 10 - 28532775 - 28583502 [+] on Build GRCh38
          GenBankmRNA DA163366.1 - 1 189 Chr. 10 - 28532768 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA070726.1 - 1 189 Chr. 10 - 28532768 - 28583476 [+] on Build GRCh38
          GenBankmRNA DA303684.1 - 1 178 Chr. 10 - 28532779 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA241683.1 - 1 183 Chr. 10 - 28532776 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA601652.1 - 1 180 Chr. 10 - 28532777 - 28583499 [+] on Build GRCh38
          GenBankmRNA DC375343.1 - 1 178 Chr. 10 - 28532779 - 28583498 [+] on Build GRCh38
          GenBankmRNA DC380490.1 - 1 207 Chr. 10 - 28532750 - 28583445 [+] on Build GRCh38
          GenBankmRNA DA405792.1 - 1 178 Chr. 10 - 28532779 - 28583500 [+] on Build GRCh38
          GenBankmRNA DA240280.1 - 1 188 Chr. 10 - 28532769 - 28535758 [+] on Build GRCh38
          GenBankmRNA DA674549.1 - 1 186 Chr. 10 - 28532773 - 28583501 [+] on Build GRCh38
          GenBankmRNA BE410391.1 - 1 174 Chr. 10 - 28532815 - 28589852 [+] on Build GRCh38
          GenBankmRNA BE385012.1 - 1 152 Chr. 10 - 28532805 - 28589845 [+] on Build GRCh38
          GenBankmRNA DC378334.1 - 1 194 Chr. 10 - 28532763 - 28583491 [+] on Build GRCh38
          GenBankmRNA DA814722.1 - 1 178 Chr. 10 - 28532779 - 28589855 [+] on Build GRCh38
          GenBankmRNA DA636108.1 - 1 185 Chr. 10 - 28532773 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA818830.1 - 1 182 Chr. 10 - 28532776 - 28583487 [+] on Build GRCh38
          GenBankmRNA DA127260.1 - 1 183 Chr. 10 - 28532775 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA067415.1 - 1 182 Chr. 10 - 28532776 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA503671.1 - 1 186 Chr. 10 - 28532773 - 28583486 [+] on Build GRCh38
          GenBankmRNA DA787593.1 - 1 183 Chr. 10 - 28532775 - 28589753 [+] on Build GRCh38
          GenBankmRNA BI819612.1 - 1 182 Chr. 10 - 28532775 - 28595771 [+] on Build GRCh38
          GenBankmRNA DA921685.1 - 1 185 Chr. 10 - 28532773 - 28583502 [+] on Build GRCh38
          GenBankmRNA DA235816.1 - 1 183 Chr. 10 - 28532776 - 28583502 [+] on Build GRCh38
          GenBankmRNA AA312024.1 - 1 154 Chr. 10 - 28532802 - 28535758 [+] on Build GRCh38
          GenBankmRNA DA727830.1 - 1 182 Chr. 10 - 28532775 - 28589842 [+] on Build GRCh38
          GenBankmRNA DA022843.1 - 1 182 Chr. 10 - 28532776 - 28583502 [+] on Build GRCh38
          GenBankmRNA DA520863.1 - 1 183 Chr. 10 - 28532776 - 28583489 [+] on Build GRCh38
          GenBankmRNA DA764589.1 - 1 183 Chr. 10 - 28532774 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA632121.1 - 1 183 Chr. 10 - 28532776 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA218926.1 - 1 185 Chr. 10 - 28532772 - 28589819 [+] on Build GRCh38
          GenBankmRNA DA831565.1 - 1 182 Chr. 10 - 28532775 - 28589836 [+] on Build GRCh38
          GenBankmRNA DC344976.1 - 1 185 Chr. 10 - 28532773 - 28535726 [+] on Build GRCh38
          GenBankmRNA DA239556.1 - 1 95 Chr. 10 - 28532862 - 28589840 [+] on Build GRCh38
          GenBankmRNA DA071119.1 - 1 182 Chr. 10 - 28532775 - 28583499 [+] on Build GRCh38
          GenBankmRNA DA204057.1 - 1 187 Chr. 10 - 28532773 - 28583502 [+] on Build GRCh38
          GenBankmRNA DA619428.1 - 1 182 Chr. 10 - 28532776 - 28589757 [+] on Build GRCh38
          GenBankmRNA DA298367.1 - 1 182 Chr. 10 - 28532775 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA289189.1 - 1 178 Chr. 10 - 28532779 - 28583475 [+] on Build GRCh38
          GenBankmRNA BE251984.1 - 1 142 Chr. 10 - 28532815 - 28610746 [+] on Build GRCh38
          GenBankmRNA DA816431.1 - 1 182 Chr. 10 - 28532775 - 28583503 [+] on Build GRCh38
          GenBankmRNA DC383856.1 - 1 186 Chr. 10 - 28532773 - 28583481 [+] on Build GRCh38
          GenBankmRNA DA626722.1 - 1 186 Chr. 10 - 28532771 - 28583482 [+] on Build GRCh38
          GenBankmRNA DA589351.1 - 1 182 Chr. 10 - 28532776 - 28583467 [+] on Build GRCh38
          GenBankmRNA DA245409.1 - 1 186 Chr. 10 - 28532773 - 28583505 [+] on Build GRCh38
          GenBankmRNA DC387781.1 - 1 182 Chr. 10 - 28532775 - 28583484 [+] on Build GRCh38
          GenBankmRNA DB075955.1 - 1 182 Chr. 10 - 28532775 - 28583481 [+] on Build GRCh38
          GenBankmRNA DC384723.1 - 1 182 Chr. 10 - 28532775 - 28583466 [+] on Build GRCh38
          GenBankmRNA DC305055.1 - 1 182 Chr. 10 - 28532775 - 28583490 [+] on Build GRCh38
          GenBankmRNA DA116252.1 - 1 182 Chr. 10 - 28532775 - 28583505 [+] on Build GRCh38
          GenBankmRNA DC310340.1 - 1 183 Chr. 10 - 28532774 - 28583483 [+] on Build GRCh38
          GenBankmRNA DA626966.1 - 1 186 Chr. 10 - 28532771 - 28583469 [+] on Build GRCh38
          GenBankmRNA DA270289.1 - 1 187 Chr. 10 - 28532773 - 28583504 [+] on Build GRCh38
          GenBankmRNA BM748830.1 - 1 64 Chr. 10 - 28532907 - 28589789 [+] on Build GRCh38
          GenBankmRNA DA248664.1 - 1 188 Chr. 10 - 28532773 - 28548060 [+] on Build GRCh38
          GenBankmRNA DA511974.1 - 1 184 Chr. 10 - 28532774 - 28583497 [+] on Build GRCh38
          GenBankmRNA DC359204.1 - 1 185 Chr. 10 - 28532773 - 28583481 [+] on Build GRCh38
          GenBankmRNA DB276388.1 - 1 178 Chr. 10 - 28532779 - 28537657 [+] on Build GRCh38
          GenBankmRNA DA305770.1 - 1 179 Chr. 10 - 28532779 - 28583478 [+] on Build GRCh38
          GenBankmRNA BG393237.1 - 1 130 Chr. 10 - 28532827 - 28590792 [+] on Build GRCh38
          GenBankmRNA DA853099.1 - 1 185 Chr. 10 - 28532773 - 28583484 [+] on Build GRCh38
          GenBankmRNA DC382935.1 - 1 178 Chr. 10 - 28532779 - 28583444 [+] on Build GRCh38
          GenBankmRNA DA854382.1 - 1 178 Chr. 10 - 28532779 - 28589754 [+] on Build GRCh38
          GenBankmRNA DC401397.1 - 1 371 Chr. 10 - 28532588 - 28535644 [+] on Build GRCh38
          GenBankmRNA DA106165.1 - 1 182 Chr. 10 - 28532776 - 28583478 [+] on Build GRCh38
          GenBankmRNA DB193470.1 - 1 186 Chr. 10 - 28532773 - 28583503 [+] on Build GRCh38
          GenBankmRNA DA689603.1 - 1 182 Chr. 10 - 28532776 - 28583468 [+] on Build GRCh38
          GenBankmRNA DA863529.1 - 1 185 Chr. 10 - 28532773 - 28583497 [+] on Build GRCh38
          GenBankmRNA DA306506.1 - 1 178 Chr. 10 - 28532779 - 28583490 [+] on Build GRCh38
          GenBankmRNA DA527001.1 - 1 185 Chr. 10 - 28532773 - 28583504 [+] on Build GRCh38
          GenBankmRNA DA931443.1 - 1 183 Chr. 10 - 28532774 - 28535758 [+] on Build GRCh38
          GenBankmRNA DA713968.1 - 1 182 Chr. 10 - 28532775 - 28583504 [+] on Build GRCh38
          GenBankmRNA DC316350.1 - 1 204 Chr. 10 - 28532754 - 28583438 [+] on Build GRCh38
          GenBankmRNA DB174923.1 - 1 186 Chr. 10 - 28532772 - 28583491 [+] on Build GRCh38
          GenBankmRNA DA109112.1 - 1 189 Chr. 10 - 28532769 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA401558.1 - 1 182 Chr. 10 - 28532775 - 28583505 [+] on Build GRCh38
          GenBankmRNA DA390628.1 - 1 143 Chr. 10 - 28532814 - 28589785 [+] on Build GRCh38
          GenBankmRNA DA933868.1 - 1 182 Chr. 10 - 28532776 - 28583503 [+] on Build GRCh38
          GenBankmRNA DA068469.1 - 1 184 Chr. 10 - 28532774 - 28583499 [+] on Build GRCh38
          GenBankmRNA DA116251.1 - 1 182 Chr. 10 - 28532775 - 28583505 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          cellular response to DNA damage stimulus
          histone monoubiquitination
          positive regulation of macroautophagy
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          G1 DNA damage checkpoint
          positive regulation of transcription, DNA-templated
          histone H2B conserved C-terminal lysine ubiquitination
          RNA polymerase II core binding
          chromatin binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-7xtrm:80/100.66.78.85:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0