Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027840.1 | 1 | 414 | |||
GenBank mRNA | AK302546.1 | - | 1 | 414 | ||
AK316308.1 | - | 1 | 44 |