Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028288.1 | 8 | 3095 | |||
GenBank mRNA | AB035335.1 | - | 5 | 3452 | ||
AB035337.1 | - | 8 | 3098 |