Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOX2-OT SIRNA Assays ›
          Gene Symbol:
          SOX2-OT
          Gene Name:
          SOX2 overlapping transcript
          Gene Aliases:
          NCRNA00043, SOX2OT
          Chromosome Location:
          Chr.3: 181056680 - 181742228 on Build GRCh38
          UniGene ID:
          Hs.654932
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263601

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SOX2-OT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_004053.3 5 961
          NR_075089.1 4 853
          NR_075090.1 3 567
          NR_075091.1 8 976
          NR_075092.1 7 868
          NR_075093.1 5 558
          GenBank mRNA AK022826.1 - 5 749
          AL157425.1 - 3 468
          BC041898.2 - 5 757
          JN711430.1 - 4 824
          JN882275.1 - 3 538
          JQ408703.1 - 8 975
          JX110620.1 - 7 1067
          JX504683.1 - 8 1029
          KF981435.1 - 4 675
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA302115.1 - 2 438 Chr. 3 - 181699533 - 181739688 [+] on Build GRCh38
          GenBankmRNA AV718176.1 - 2 407 Chr. 3 - 181699563 - 181740003 [+] on Build GRCh38
          GenBankmRNA DA261948.1 - 2 360 Chr. 3 - 181699612 - 181739850 [+] on Build GRCh38
          GenBankmRNA DA132672.1 - 2 396 Chr. 3 - 181699575 - 181739808 [+] on Build GRCh38
          GenBankmRNA DA278319.1 - 3 491 Chr. 3 - 181699588 - 181739728 [+] on Build GRCh38
          GenBankmRNA DA133155.1 - 2 396 Chr. 3 - 181699575 - 181739805 [+] on Build GRCh38
          GenBankmRNA DA263226.1 - 3 505 Chr. 3 - 181699574 - 181739701 [+] on Build GRCh38
          GenBankmRNA DA139670.1 - 3 552 Chr. 3 - 181699527 - 181739684 [+] on Build GRCh38
          GenBankmRNA BX423294.2 - 2 355 Chr. 3 - 181699616 - 181740257 [+] on Build GRCh38
          GenBankmRNA DA282731.1 - 2 432 Chr. 3 - 181699546 - 181739781 [+] on Build GRCh38
          GenBankmRNA DA359248.1 - 3 509 Chr. 3 - 181699570 - 181739702 [+] on Build GRCh38
          GenBankmRNA DA365386.1 - 3 477 Chr. 3 - 181699604 - 181739745 [+] on Build GRCh38
          GenBankmRNA DB175041.1 - 3 504 Chr. 3 - 181699575 - 181739712 [+] on Build GRCh38
          GenBankmRNA DA179672.1 - 2 396 Chr. 3 - 181699575 - 181739782 [+] on Build GRCh38
          GenBankmRNA DB494090.1 - 2 364 Chr. 3 - 181699606 - 181739821 [+] on Build GRCh38
          GenBankmRNA DA270048.1 - 3 503 Chr. 3 - 181699576 - 181739718 [+] on Build GRCh38
          GenBankmRNA DA154575.1 - 3 466 Chr. 3 - 181699612 - 181739750 [+] on Build GRCh38
          GenBankmRNA DB175631.1 - 3 503 Chr. 3 - 181699576 - 181739715 [+] on Build GRCh38
          GenBankmRNA DA132079.1 - 3 506 Chr. 3 - 181699575 - 181739705 [+] on Build GRCh38
          GenBankmRNA DA195694.1 - 2 396 Chr. 3 - 181699575 - 181739829 [+] on Build GRCh38
          GenBankmRNA DA268973.1 - 3 504 Chr. 3 - 181699575 - 181739707 [+] on Build GRCh38
          GenBankmRNA DA142762.1 - 2 396 Chr. 3 - 181699575 - 181739817 [+] on Build GRCh38
          GenBankmRNA DA277970.1 - 3 504 Chr. 3 - 181699575 - 181739675 [+] on Build GRCh38
          GenBankmRNA DA275725.1 - 3 467 Chr. 3 - 181699612 - 181739743 [+] on Build GRCh38
          GenBankmRNA BX442540.2 - 2 372 Chr. 3 - 181699598 - 181740289 [+] on Build GRCh38
          GenBankmRNA DA284814.1 - 2 395 Chr. 3 - 181699575 - 181739803 [+] on Build GRCh38
          GenBankmRNA DA284155.1 - 3 542 Chr. 3 - 181699546 - 181739675 [+] on Build GRCh38
          GenBankmRNA DA311296.1 - 2 397 Chr. 3 - 181699575 - 181739805 [+] on Build GRCh38
          GenBankmRNA DA343465.1 - 3 473 Chr. 3 - 181699606 - 181739756 [+] on Build GRCh38
          GenBankmRNA DA165213.1 - 3 516 Chr. 3 - 181699575 - 181739701 [+] on Build GRCh38
          GenBankmRNA DA354554.1 - 3 505 Chr. 3 - 181699575 - 181739732 [+] on Build GRCh38
          GenBankmRNA DA532516.1 - 2 416 Chr. 3 - 181699559 - 181739734 [+] on Build GRCh38
          GenBankmRNA DA286721.1 - 2 396 Chr. 3 - 181699575 - 181739809 [+] on Build GRCh38
          GenBankmRNA DA201533.1 - 2 390 Chr. 3 - 181699581 - 181739839 [+] on Build GRCh38
          GenBankmRNA DA418095.1 - 2 406 Chr. 3 - 181699575 - 181739798 [+] on Build GRCh38
          GenBankmRNA DA358810.1 - 2 396 Chr. 3 - 181699575 - 181739836 [+] on Build GRCh38
          GenBankmRNA DA278267.1 - 3 504 Chr. 3 - 181699575 - 181739709 [+] on Build GRCh38
          GenBankmRNA DA180052.1 - 3 504 Chr. 3 - 181699575 - 181739720 [+] on Build GRCh38
          GenBankmRNA DA148233.1 - 3 510 Chr. 3 - 181699570 - 181739725 [+] on Build GRCh38
          GenBankmRNA DA136099.1 - 3 504 Chr. 3 - 181699575 - 181739785 [+] on Build GRCh38
          GenBankmRNA DA140256.1 - 3 492 Chr. 3 - 181699587 - 181739722 [+] on Build GRCh38
          GenBankmRNA DB480926.1 - 2 364 Chr. 3 - 181699607 - 181739765 [+] on Build GRCh38
          GenBankmRNA HY111336.1 - 2 368 Chr. 3 - 181699602 - 181739815 [+] on Build GRCh38
          GenBankmRNA DA349471.1 - 3 522 Chr. 3 - 181699559 - 181739691 [+] on Build GRCh38
          GenBankmRNA DA269055.1 - 2 155 Chr. 3 - 181715224 - 181740038 [+] on Build GRCh38
          GenBankmRNA DA265083.1 - 2 427 Chr. 3 - 181699546 - 181739764 [+] on Build GRCh38
          GenBankmRNA DA368208.1 - 3 502 Chr. 3 - 181699576 - 181739737 [+] on Build GRCh38
          GenBankmRNA DA406174.1 - 2 396 Chr. 3 - 181699575 - 181739810 [+] on Build GRCh38
          GenBankmRNA DA153050.1 - 2 398 Chr. 3 - 181699575 - 181739830 [+] on Build GRCh38
          GenBankmRNA AV725391.1 - 3 513 Chr. 3 - 181699565 - 181739947 [+] on Build GRCh38
          GenBankmRNA HY130776.1 - 2 368 Chr. 3 - 181699602 - 181739757 [+] on Build GRCh38
          GenBankmRNA DA355176.1 - 2 396 Chr. 3 - 181699575 - 181739806 [+] on Build GRCh38
          GenBankmRNA DA412243.1 - 3 503 Chr. 3 - 181699575 - 181739732 [+] on Build GRCh38
          GenBankmRNA DA284596.1 - 2 397 Chr. 3 - 181699575 - 181739764 [+] on Build GRCh38
          GenBankmRNA BX459910.2 - 2 354 Chr. 3 - 181699616 - 181740148 [+] on Build GRCh38
          GenBankmRNA BG701492.1 - 3 510 Chr. 3 - 181699575 - 181739974 [+] on Build GRCh38
          GenBankmRNA DA136383.1 - 2 441 Chr. 3 - 181699529 - 181739759 [+] on Build GRCh38
          GenBankmRNA DA280038.1 - 2 397 Chr. 3 - 181699575 - 181739827 [+] on Build GRCh38
          GenBankmRNA DB180169.1 - 3 505 Chr. 3 - 181699575 - 181739704 [+] on Build GRCh38
          GenBankmRNA DA408016.1 - 2 395 Chr. 3 - 181699575 - 181739804 [+] on Build GRCh38
          GenBankmRNA DA352576.1 - 3 503 Chr. 3 - 181699575 - 181739732 [+] on Build GRCh38
          GenBankmRNA DA358441.1 - 2 409 Chr. 3 - 181699575 - 181739790 [+] on Build GRCh38
          GenBankmRNA HY110405.1 - 2 252 Chr. 3 - 181699719 - 181739927 [+] on Build GRCh38
          GenBankmRNA BI601220.1 - 3 506 Chr. 3 - 181699575 - 181739843 [+] on Build GRCh38
          GenBankmRNA DA139532.1 - 2 395 Chr. 3 - 181699575 - 181739884 [+] on Build GRCh38
          GenBankmRNA DA362847.1 - 2 396 Chr. 3 - 181699575 - 181739822 [+] on Build GRCh38
          GenBankmRNA DA735954.1 - 4 663 Chr. 3 - 181610524 - 181739880 [+] on Build GRCh38
          GenBankmRNA DB169465.1 - 3 504 Chr. 3 - 181699575 - 181739741 [+] on Build GRCh38
          GenBankmRNA DB170935.1 - 2 452 Chr. 3 - 181699518 - 181739774 [+] on Build GRCh38
          GenBankmRNA DA270634.1 - 2 387 Chr. 3 - 181699584 - 181739812 [+] on Build GRCh38
          GenBankmRNA DA189377.1 - 2 395 Chr. 3 - 181699576 - 181739855 [+] on Build GRCh38
          GenBankmRNA DA284723.1 - 2 396 Chr. 3 - 181699576 - 181739810 [+] on Build GRCh38
          GenBankmRNA DA254884.1 - 3 312 Chr. 3 - 181699766 - 181739903 [+] on Build GRCh38
          GenBankmRNA DA131719.1 - 2 396 Chr. 3 - 181699575 - 181739799 [+] on Build GRCh38
          GenBankmRNA DA286610.1 - 3 504 Chr. 3 - 181699575 - 181739727 [+] on Build GRCh38
          GenBankmRNA DA260132.1 - 2 397 Chr. 3 - 181699576 - 181739821 [+] on Build GRCh38
          GenBankmRNA DA129372.1 - 2 395 Chr. 3 - 181699575 - 181739828 [+] on Build GRCh38
          GenBankmRNA DA131865.1 - 3 511 Chr. 3 - 181699569 - 181739685 [+] on Build GRCh38
          GenBankmRNA DA345227.1 - 2 365 Chr. 3 - 181699606 - 181739855 [+] on Build GRCh38
          GenBankmRNA BG715434.1 - 2 400 Chr. 3 - 181699575 - 181739875 [+] on Build GRCh38
          GenBankmRNA DA277929.1 - 2 477 Chr. 3 - 181699493 - 181739737 [+] on Build GRCh38
          GenBankmRNA DA167857.1 - 3 504 Chr. 3 - 181699575 - 181739728 [+] on Build GRCh38
          GenBankmRNA DA175390.1 - 3 547 Chr. 3 - 181699531 - 181739676 [+] on Build GRCh38
          GenBankmRNA DA274461.1 - 2 396 Chr. 3 - 181699575 - 181739832 [+] on Build GRCh38
          GenBankmRNA DA281737.1 - 2 395 Chr. 3 - 181699575 - 181739827 [+] on Build GRCh38
          GenBankmRNA DA272665.1 - 3 503 Chr. 3 - 181699575 - 181739684 [+] on Build GRCh38
          GenBankmRNA DB495259.1 - 2 364 Chr. 3 - 181699606 - 181739780 [+] on Build GRCh38
          GenBankmRNA DA287748.1 - 2 396 Chr. 3 - 181699575 - 181739810 [+] on Build GRCh38
          GenBankmRNA DA350535.1 - 2 395 Chr. 3 - 181699575 - 181739849 [+] on Build GRCh38
          GenBankmRNA BI545922.1 - 3 551 Chr. 3 - 181699533 - 181739922 [+] on Build GRCh38
          GenBankmRNA DA104936.1 - 2 395 Chr. 3 - 181699577 - 181739848 [+] on Build GRCh38
          GenBankmRNA DA369884.1 - 3 491 Chr. 3 - 181699588 - 181739716 [+] on Build GRCh38
          GenBankmRNA DA166395.1 - 2 395 Chr. 3 - 181699575 - 181739785 [+] on Build GRCh38
          GenBankmRNA DA279068.1 - 3 504 Chr. 3 - 181699575 - 181739717 [+] on Build GRCh38
          GenBankmRNA DA142203.1 - 3 467 Chr. 3 - 181699612 - 181739740 [+] on Build GRCh38
          GenBankmRNA DA208447.1 - 3 545 Chr. 3 - 181699533 - 181739683 [+] on Build GRCh38
          GenBankmRNA DA133120.1 - 2 396 Chr. 3 - 181699575 - 181739806 [+] on Build GRCh38
          GenBankmRNA DA350278.1 - 3 503 Chr. 3 - 181699575 - 181739675 [+] on Build GRCh38
          GenBankmRNA DB178263.1 - 2 395 Chr. 3 - 181699575 - 181739814 [+] on Build GRCh38
          GenBankmRNA DB172455.1 - 2 396 Chr. 3 - 181699575 - 181739820 [+] on Build GRCh38
          GenBankmRNA DA290999.1 - 3 505 Chr. 3 - 181699575 - 181739724 [+] on Build GRCh38
          GenBankmRNA DT219358.1 - 3 503 Chr. 3 - 181699575 - 181739818 [+] on Build GRCh38
          GenBankmRNA DA357302.1 - 3 503 Chr. 3 - 181699575 - 181739702 [+] on Build GRCh38
          GenBankmRNA DA134901.1 - 3 504 Chr. 3 - 181699575 - 181739706 [+] on Build GRCh38
          GenBankmRNA DA369506.1 - 3 509 Chr. 3 - 181699570 - 181739718 [+] on Build GRCh38
          GenBankmRNA DA248954.1 - 3 504 Chr. 3 - 181699575 - 181739729 [+] on Build GRCh38
          GenBankmRNA DA362148.1 - 3 503 Chr. 3 - 181699575 - 181739729 [+] on Build GRCh38
          GenBankmRNA DA233431.1 - 3 504 Chr. 3 - 181699575 - 181739727 [+] on Build GRCh38
          GenBankmRNA BI601514.1 - 3 506 Chr. 3 - 181699575 - 181739998 [+] on Build GRCh38
          GenBankmRNA DA277798.1 - 2 367 Chr. 3 - 181699612 - 181739778 [+] on Build GRCh38
          GenBankmRNA DA278757.1 - 2 398 Chr. 3 - 181699577 - 181739805 [+] on Build GRCh38
          GenBankmRNA DA362437.1 - 2 396 Chr. 3 - 181699575 - 181739729 [+] on Build GRCh38
          GenBankmRNA DA346811.1 - 3 505 Chr. 3 - 181699577 - 181739706 [+] on Build GRCh38
          GenBankmRNA DA308672.1 - 3 538 Chr. 3 - 181610363 - 181739693 [+] on Build GRCh38
          GenBankmRNA BI549819.1 - 2 385 Chr. 3 - 181699589 - 181740014 [+] on Build GRCh38
          GenBankmRNA DB175439.1 - 2 428 Chr. 3 - 181699546 - 181739816 [+] on Build GRCh38
          GenBankmRNA DA352078.1 - 2 395 Chr. 3 - 181699575 - 181739818 [+] on Build GRCh38
          GenBankmRNA DA160000.1 - 2 397 Chr. 3 - 181699575 - 181739815 [+] on Build GRCh38
          GenBankmRNA DA277074.1 - 3 506 Chr. 3 - 181699575 - 181739726 [+] on Build GRCh38
          GenBankmRNA DA531399.1 - 2 396 Chr. 3 - 181699576 - 181739876 [+] on Build GRCh38
          GenBankmRNA DA285311.1 - 3 504 Chr. 3 - 181699575 - 181739689 [+] on Build GRCh38
          GenBankmRNA DA303463.1 - 3 505 Chr. 3 - 181699577 - 181739737 [+] on Build GRCh38
          GenBankmRNA DA270443.1 - 3 475 Chr. 3 - 181699604 - 181739755 [+] on Build GRCh38
          GenBankmRNA DA363633.1 - 2 407 Chr. 3 - 181699575 - 181739821 [+] on Build GRCh38
          GenBankmRNA DA283659.1 - 3 483 Chr. 3 - 181699596 - 181739753 [+] on Build GRCh38
          GenBankmRNA DA181278.1 - 2 383 Chr. 3 - 181699587 - 181739839 [+] on Build GRCh38
          GenBankmRNA DA168440.1 - 2 396 Chr. 3 - 181699575 - 181739847 [+] on Build GRCh38
          GenBankmRNA BM725940.1 - 2 397 Chr. 3 - 181699573 - 181739882 [+] on Build GRCh38
          GenBankmRNA DA367963.1 - 3 503 Chr. 3 - 181699575 - 181739710 [+] on Build GRCh38
          GenBankmRNA DA279363.1 - 2 395 Chr. 3 - 181699575 - 181739844 [+] on Build GRCh38
          GenBankmRNA DA529805.1 - 2 402 Chr. 3 - 181699575 - 181739860 [+] on Build GRCh38
          GenBankmRNA DA345402.1 - 3 504 Chr. 3 - 181699575 - 181739714 [+] on Build GRCh38
          GenBankmRNA DA145889.1 - 2 398 Chr. 3 - 181699575 - 181739823 [+] on Build GRCh38
          GenBankmRNA DA342800.1 - 3 503 Chr. 3 - 181699575 - 181739728 [+] on Build GRCh38
          GenBankmRNA DA357231.1 - 3 467 Chr. 3 - 181699612 - 181739753 [+] on Build GRCh38
          GenBankmRNA DA134835.1 - 2 386 Chr. 3 - 181699585 - 181739838 [+] on Build GRCh38
          GenBankmRNA DA286657.1 - 3 534 Chr. 3 - 181699546 - 181739688 [+] on Build GRCh38
          GenBankmRNA DB474842.1 - 3 505 Chr. 3 - 181699575 - 181739664 [+] on Build GRCh38
          GenBankmRNA BI757585.1 - 2 457 Chr. 3 - 181699515 - 181740032 [+] on Build GRCh38
          GenBankmRNA DA159447.1 - 3 504 Chr. 3 - 181699575 - 181739721 [+] on Build GRCh38
          GenBankmRNA DA284362.1 - 2 395 Chr. 3 - 181699575 - 181739749 [+] on Build GRCh38
          GenBankmRNA DA406068.1 - 3 504 Chr. 3 - 181699576 - 181739736 [+] on Build GRCh38
          GenBankmRNA DA166593.1 - 3 509 Chr. 3 - 181699577 - 181739690 [+] on Build GRCh38
          GenBankmRNA DB173817.1 - 2 414 Chr. 3 - 181699559 - 181739811 [+] on Build GRCh38
          GenBankmRNA DA351387.1 - 3 506 Chr. 3 - 181699575 - 181739739 [+] on Build GRCh38
          GenBankmRNA DA264513.1 - 2 396 Chr. 3 - 181699575 - 181739815 [+] on Build GRCh38
          GenBankmRNA DA273021.1 - 2 396 Chr. 3 - 181699575 - 181739838 [+] on Build GRCh38
          GenBankmRNA BI916139.1 - 3 551 Chr. 3 - 181699526 - 181739917 [+] on Build GRCh38
          GenBankmRNA BI668091.1 - 3 503 Chr. 3 - 181699598 - 181739968 [+] on Build GRCh38
          GenBankmRNA DA298250.1 - 3 467 Chr. 3 - 181699612 - 181739750 [+] on Build GRCh38
          GenBankmRNA HY126742.1 - 2 373 Chr. 3 - 181699597 - 181739750 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0