Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOX2-OT SIRNA Assays ›
          Gene Symbol:
          SOX2-OT
          Gene Name:
          SOX2 overlapping transcript
          Gene Aliases:
          NCRNA00043, SOX2OT
          Chromosome Location:
          Chr.3: 181056680 - 181742228 on Build GRCh38
          UniGene ID:
          Hs.654932
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263602

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SOX2-OT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_004053.3 1 381
          NR_075089.1 1 381
          NR_075090.1 1 381
          GenBank mRNA AK022826.1 - 1 169
          DA250340.1 - 1 352
          DA366949.1 - 1 381
          JN711430.1 - 1 352
          JN882275.1 - 1 352
          KF981435.1 - 1 381
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AU127535.1 - 1 169 Chr. 3 - 181610546 - 181739643 [+] on Build GRCh38
          GenBankmRNA BM474966.1 - 1 106 Chr. 3 - 181610609 - 181710237 [+] on Build GRCh38
          GenBankmRNA DA308672.1 - 1 352 Chr. 3 - 181610363 - 181739693 [+] on Build GRCh38
          GenBankmRNA DB170062.1 - 1 186 Chr. 3 - 181610530 - 181699885 [+] on Build GRCh38
          GenBankmRNA DA175606.1 - 1 130 Chr. 3 - 181610587 - 181699885 [+] on Build GRCh38
          GenBankmRNA BI600313.1 - 1 216 Chr. 3 - 181610500 - 181739601 [+] on Build GRCh38
          GenBankmRNA DA133714.1 - 1 186 Chr. 3 - 181610529 - 181790946 [+] on Build GRCh38
          GenBankmRNA DA485580.1 - 1 191 Chr. 3 - 181610524 - 181699883 [+] on Build GRCh38
          GenBankmRNA BX110135.1 - 1 75 Chr. 3 - 181610640 - 181836475 [+] on Build GRCh38
          GenBankmRNA DA353700.1 - 1 129 Chr. 3 - 181610586 - 181715225 [+] on Build GRCh38
          GenBankmRNA DA731984.1 - 1 191 Chr. 3 - 181610524 - 181699769 [+] on Build GRCh38
          GenBankmRNA DA279403.1 - 1 197 Chr. 3 - 181610518 - 181699880 [+] on Build GRCh38
          GenBankmRNA DA735954.1 - 1 191 Chr. 3 - 181610524 - 181739880 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0