Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOX2-OT SIRNA Assays ›
          Gene Symbol:
          SOX2-OT
          Gene Name:
          SOX2 overlapping transcript
          Gene Aliases:
          NCRNA00043, SOX2OT
          Chromosome Location:
          Chr.3: 181056680 - 181742228 on Build GRCh38
          UniGene ID:
          Hs.654932
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263605

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SOX2-OT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_004053.3 4, 5 886
          NR_075089.1 3, 4 778
          NR_075090.1 2, 3 492
          NR_075091.1 7, 8 901
          NR_075092.1 6, 7 793
          NR_075093.1 4, 5 483
          GenBank mRNA AK022826.1 - 4, 5 674
          AL157425.1 - 2, 3 393
          BC041898.2 - 4, 5 682
          DA310380.1 - 4, 5 483
          DA366949.1 - 2, 3 492
          JN711430.1 - 3, 4 749
          JN882275.1 - 2, 3 463
          JQ408703.1 - 7, 8 900
          JX110620.1 - 6, 7 992
          JX504683.1 - 7, 8 954
          KF981435.1 - 3, 4 600
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA302115.1 - 1, 2 363 Chr. 3 - 181699533 - 181739688 [+] on Build GRCh38
          GenBankmRNA AV718176.1 - 1, 2 332 Chr. 3 - 181699563 - 181740003 [+] on Build GRCh38
          GenBankmRNA DA261948.1 - 1, 2 285 Chr. 3 - 181699612 - 181739850 [+] on Build GRCh38
          GenBankmRNA DA132672.1 - 1, 2 321 Chr. 3 - 181699575 - 181739808 [+] on Build GRCh38
          GenBankmRNA DA278319.1 - 2, 3 416 Chr. 3 - 181699588 - 181739728 [+] on Build GRCh38
          GenBankmRNA DA263226.1 - 2, 3 430 Chr. 3 - 181699574 - 181739701 [+] on Build GRCh38
          GenBankmRNA DA133155.1 - 1, 2 321 Chr. 3 - 181699575 - 181739805 [+] on Build GRCh38
          GenBankmRNA DA139670.1 - 2, 3 477 Chr. 3 - 181699527 - 181739684 [+] on Build GRCh38
          GenBankmRNA DA282731.1 - 1, 2 357 Chr. 3 - 181699546 - 181739781 [+] on Build GRCh38
          GenBankmRNA BX423294.2 - 1, 2 280 Chr. 3 - 181699616 - 181740257 [+] on Build GRCh38
          GenBankmRNA DB175041.1 - 2, 3 429 Chr. 3 - 181699575 - 181739712 [+] on Build GRCh38
          GenBankmRNA DA359248.1 - 2, 3 434 Chr. 3 - 181699570 - 181739702 [+] on Build GRCh38
          GenBankmRNA DA365386.1 - 2, 3 402 Chr. 3 - 181699604 - 181739745 [+] on Build GRCh38
          GenBankmRNA DA179672.1 - 1, 2 321 Chr. 3 - 181699575 - 181739782 [+] on Build GRCh38
          GenBankmRNA DB494090.1 - 1, 2 289 Chr. 3 - 181699606 - 181739821 [+] on Build GRCh38
          GenBankmRNA DA132079.1 - 2, 3 431 Chr. 3 - 181699575 - 181739705 [+] on Build GRCh38
          GenBankmRNA DA270048.1 - 2, 3 428 Chr. 3 - 181699576 - 181739718 [+] on Build GRCh38
          GenBankmRNA DB175631.1 - 2, 3 428 Chr. 3 - 181699576 - 181739715 [+] on Build GRCh38
          GenBankmRNA DA154575.1 - 2, 3 391 Chr. 3 - 181699612 - 181739750 [+] on Build GRCh38
          GenBankmRNA DA195694.1 - 1, 2 321 Chr. 3 - 181699575 - 181739829 [+] on Build GRCh38
          GenBankmRNA DA268973.1 - 2, 3 429 Chr. 3 - 181699575 - 181739707 [+] on Build GRCh38
          GenBankmRNA DA142762.1 - 1, 2 321 Chr. 3 - 181699575 - 181739817 [+] on Build GRCh38
          GenBankmRNA DA277970.1 - 2, 3 429 Chr. 3 - 181699575 - 181739675 [+] on Build GRCh38
          GenBankmRNA DA275725.1 - 2, 3 392 Chr. 3 - 181699612 - 181739743 [+] on Build GRCh38
          GenBankmRNA BX442540.2 - 1, 2 297 Chr. 3 - 181699598 - 181740289 [+] on Build GRCh38
          GenBankmRNA DA284155.1 - 2, 3 467 Chr. 3 - 181699546 - 181739675 [+] on Build GRCh38
          GenBankmRNA DA311296.1 - 1, 2 322 Chr. 3 - 181699575 - 181739805 [+] on Build GRCh38
          GenBankmRNA DA343465.1 - 2, 3 398 Chr. 3 - 181699606 - 181739756 [+] on Build GRCh38
          GenBankmRNA DA165213.1 - 2, 3 441 Chr. 3 - 181699575 - 181739701 [+] on Build GRCh38
          GenBankmRNA DA354554.1 - 2, 3 430 Chr. 3 - 181699575 - 181739732 [+] on Build GRCh38
          GenBankmRNA DA286721.1 - 1, 2 321 Chr. 3 - 181699575 - 181739809 [+] on Build GRCh38
          GenBankmRNA DA532516.1 - 1, 2 341 Chr. 3 - 181699559 - 181739734 [+] on Build GRCh38
          GenBankmRNA DA201533.1 - 1, 2 315 Chr. 3 - 181699581 - 181739839 [+] on Build GRCh38
          GenBankmRNA DA418095.1 - 1, 2 331 Chr. 3 - 181699575 - 181739798 [+] on Build GRCh38
          GenBankmRNA DA358810.1 - 1, 2 321 Chr. 3 - 181699575 - 181739836 [+] on Build GRCh38
          GenBankmRNA DA278267.1 - 2, 3 429 Chr. 3 - 181699575 - 181739709 [+] on Build GRCh38
          GenBankmRNA DA180052.1 - 2, 3 429 Chr. 3 - 181699575 - 181739720 [+] on Build GRCh38
          GenBankmRNA DA148233.1 - 2, 3 435 Chr. 3 - 181699570 - 181739725 [+] on Build GRCh38
          GenBankmRNA DA136099.1 - 2, 3 429 Chr. 3 - 181699575 - 181739785 [+] on Build GRCh38
          GenBankmRNA DA140256.1 - 2, 3 417 Chr. 3 - 181699587 - 181739722 [+] on Build GRCh38
          GenBankmRNA HY111336.1 - 1, 2 293 Chr. 3 - 181699602 - 181739815 [+] on Build GRCh38
          GenBankmRNA DB480926.1 - 1, 2 289 Chr. 3 - 181699607 - 181739765 [+] on Build GRCh38
          GenBankmRNA DA349471.1 - 2, 3 447 Chr. 3 - 181699559 - 181739691 [+] on Build GRCh38
          GenBankmRNA DB505934.1 - 2, 3 428 Chr. 3 - 181699575 - 181739658 [+] on Build GRCh38
          GenBankmRNA DA269055.1 - 1, 2 80 Chr. 3 - 181715224 - 181740038 [+] on Build GRCh38
          GenBankmRNA DA265083.1 - 1, 2 352 Chr. 3 - 181699546 - 181739764 [+] on Build GRCh38
          GenBankmRNA DA368208.1 - 2, 3 427 Chr. 3 - 181699576 - 181739737 [+] on Build GRCh38
          GenBankmRNA DA406174.1 - 1, 2 321 Chr. 3 - 181699575 - 181739810 [+] on Build GRCh38
          GenBankmRNA DA153050.1 - 1, 2 323 Chr. 3 - 181699575 - 181739830 [+] on Build GRCh38
          GenBankmRNA AV725391.1 - 2, 3 438 Chr. 3 - 181699565 - 181739947 [+] on Build GRCh38
          GenBankmRNA HY130776.1 - 1, 2 293 Chr. 3 - 181699602 - 181739757 [+] on Build GRCh38
          GenBankmRNA DA412243.1 - 2, 3 428 Chr. 3 - 181699575 - 181739732 [+] on Build GRCh38
          GenBankmRNA DA355176.1 - 1, 2 321 Chr. 3 - 181699575 - 181739806 [+] on Build GRCh38
          GenBankmRNA DA284596.1 - 1, 2 322 Chr. 3 - 181699575 - 181739764 [+] on Build GRCh38
          GenBankmRNA BX459910.2 - 1, 2 279 Chr. 3 - 181699616 - 181740148 [+] on Build GRCh38
          GenBankmRNA BG701492.1 - 2, 3 435 Chr. 3 - 181699575 - 181739974 [+] on Build GRCh38
          GenBankmRNA DA136383.1 - 1, 2 366 Chr. 3 - 181699529 - 181739759 [+] on Build GRCh38
          GenBankmRNA DA259497.1 - 2, 3 428 Chr. 3 - 181699576 - 181739615 [+] on Build GRCh38
          GenBankmRNA DA280038.1 - 1, 2 322 Chr. 3 - 181699575 - 181739827 [+] on Build GRCh38
          GenBankmRNA DB180169.1 - 2, 3 430 Chr. 3 - 181699575 - 181739704 [+] on Build GRCh38
          GenBankmRNA DA408016.1 - 1, 2 320 Chr. 3 - 181699575 - 181739804 [+] on Build GRCh38
          GenBankmRNA DA352576.1 - 2, 3 428 Chr. 3 - 181699575 - 181739732 [+] on Build GRCh38
          GenBankmRNA DA358441.1 - 1, 2 334 Chr. 3 - 181699575 - 181739790 [+] on Build GRCh38
          GenBankmRNA HY110405.1 - 1, 2 177 Chr. 3 - 181699719 - 181739927 [+] on Build GRCh38
          GenBankmRNA BI601220.1 - 2, 3 431 Chr. 3 - 181699575 - 181739843 [+] on Build GRCh38
          GenBankmRNA DA139532.1 - 1, 2 320 Chr. 3 - 181699575 - 181739884 [+] on Build GRCh38
          GenBankmRNA DA362847.1 - 1, 2 321 Chr. 3 - 181699575 - 181739822 [+] on Build GRCh38
          GenBankmRNA DA735954.1 - 3, 4 588 Chr. 3 - 181610524 - 181739880 [+] on Build GRCh38
          GenBankmRNA DB169465.1 - 2, 3 429 Chr. 3 - 181699575 - 181739741 [+] on Build GRCh38
          GenBankmRNA DB170935.1 - 1, 2 377 Chr. 3 - 181699518 - 181739774 [+] on Build GRCh38
          GenBankmRNA DA270634.1 - 1, 2 312 Chr. 3 - 181699584 - 181739812 [+] on Build GRCh38
          GenBankmRNA DA189377.1 - 1, 2 320 Chr. 3 - 181699576 - 181739855 [+] on Build GRCh38
          GenBankmRNA DA284723.1 - 1, 2 321 Chr. 3 - 181699576 - 181739810 [+] on Build GRCh38
          GenBankmRNA DA131719.1 - 1, 2 321 Chr. 3 - 181699575 - 181739799 [+] on Build GRCh38
          GenBankmRNA DA254884.1 - 2, 3 237 Chr. 3 - 181699766 - 181739903 [+] on Build GRCh38
          GenBankmRNA DB480681.1 - 2, 3 425 Chr. 3 - 181699578 - 181739624 [+] on Build GRCh38
          GenBankmRNA DA164011.1 - 2, 3 509 Chr. 3 - 181699496 - 181739642 [+] on Build GRCh38
          GenBankmRNA DA286610.1 - 2, 3 429 Chr. 3 - 181699575 - 181739727 [+] on Build GRCh38
          GenBankmRNA DA260132.1 - 1, 2 322 Chr. 3 - 181699576 - 181739821 [+] on Build GRCh38
          GenBankmRNA BI600313.1 - 4, 5 722 Chr. 3 - 181610500 - 181739601 [+] on Build GRCh38
          GenBankmRNA DA129372.1 - 1, 2 320 Chr. 3 - 181699575 - 181739828 [+] on Build GRCh38
          GenBankmRNA DA345227.1 - 1, 2 290 Chr. 3 - 181699606 - 181739855 [+] on Build GRCh38
          GenBankmRNA BG715434.1 - 1, 2 325 Chr. 3 - 181699575 - 181739875 [+] on Build GRCh38
          GenBankmRNA DA131865.1 - 2, 3 436 Chr. 3 - 181699569 - 181739685 [+] on Build GRCh38
          GenBankmRNA DA277929.1 - 1, 2 402 Chr. 3 - 181699493 - 181739737 [+] on Build GRCh38
          GenBankmRNA DA167857.1 - 2, 3 429 Chr. 3 - 181699575 - 181739728 [+] on Build GRCh38
          GenBankmRNA DA175390.1 - 2, 3 472 Chr. 3 - 181699531 - 181739676 [+] on Build GRCh38
          GenBankmRNA DA362189.1 - 2, 3 435 Chr. 3 - 181699575 - 181739608 [+] on Build GRCh38
          GenBankmRNA DA281737.1 - 1, 2 320 Chr. 3 - 181699575 - 181739827 [+] on Build GRCh38
          GenBankmRNA DA274461.1 - 1, 2 321 Chr. 3 - 181699575 - 181739832 [+] on Build GRCh38
          GenBankmRNA DA257051.1 - 2, 3 428 Chr. 3 - 181699575 - 181739615 [+] on Build GRCh38
          GenBankmRNA DA272665.1 - 2, 3 428 Chr. 3 - 181699575 - 181739684 [+] on Build GRCh38
          GenBankmRNA DA287748.1 - 1, 2 321 Chr. 3 - 181699575 - 181739810 [+] on Build GRCh38
          GenBankmRNA DA350535.1 - 1, 2 320 Chr. 3 - 181699575 - 181739849 [+] on Build GRCh38
          GenBankmRNA DA104936.1 - 1, 2 320 Chr. 3 - 181699577 - 181739848 [+] on Build GRCh38
          GenBankmRNA DA264487.1 - 2, 3 429 Chr. 3 - 181699575 - 181739615 [+] on Build GRCh38
          GenBankmRNA DA166395.1 - 1, 2 320 Chr. 3 - 181699575 - 181739785 [+] on Build GRCh38
          GenBankmRNA DA369884.1 - 2, 3 416 Chr. 3 - 181699588 - 181739716 [+] on Build GRCh38
          GenBankmRNA DA279068.1 - 2, 3 429 Chr. 3 - 181699575 - 181739717 [+] on Build GRCh38
          GenBankmRNA DB482709.1 - 2, 3 430 Chr. 3 - 181699575 - 181739650 [+] on Build GRCh38
          GenBankmRNA DA142203.1 - 2, 3 392 Chr. 3 - 181699612 - 181739740 [+] on Build GRCh38
          GenBankmRNA DA208447.1 - 2, 3 470 Chr. 3 - 181699533 - 181739683 [+] on Build GRCh38
          GenBankmRNA DA133120.1 - 1, 2 321 Chr. 3 - 181699575 - 181739806 [+] on Build GRCh38
          GenBankmRNA DA350278.1 - 2, 3 428 Chr. 3 - 181699575 - 181739675 [+] on Build GRCh38
          GenBankmRNA DA130431.1 - 2, 3 476 Chr. 3 - 181699527 - 181739662 [+] on Build GRCh38
          GenBankmRNA DB178263.1 - 1, 2 320 Chr. 3 - 181699575 - 181739814 [+] on Build GRCh38
          GenBankmRNA DA290999.1 - 2, 3 430 Chr. 3 - 181699575 - 181739724 [+] on Build GRCh38
          GenBankmRNA DB172455.1 - 1, 2 321 Chr. 3 - 181699575 - 181739820 [+] on Build GRCh38
          GenBankmRNA DT219358.1 - 2, 3 428 Chr. 3 - 181699575 - 181739818 [+] on Build GRCh38
          GenBankmRNA DA357302.1 - 2, 3 428 Chr. 3 - 181699575 - 181739702 [+] on Build GRCh38
          GenBankmRNA DA134901.1 - 2, 3 429 Chr. 3 - 181699575 - 181739706 [+] on Build GRCh38
          GenBankmRNA DA369506.1 - 2, 3 434 Chr. 3 - 181699570 - 181739718 [+] on Build GRCh38
          GenBankmRNA DA248954.1 - 2, 3 429 Chr. 3 - 181699575 - 181739729 [+] on Build GRCh38
          GenBankmRNA DA290283.1 - 2, 3 503 Chr. 3 - 181699500 - 181739658 [+] on Build GRCh38
          GenBankmRNA DA362148.1 - 2, 3 428 Chr. 3 - 181699575 - 181739729 [+] on Build GRCh38
          GenBankmRNA DA233431.1 - 2, 3 429 Chr. 3 - 181699575 - 181739727 [+] on Build GRCh38
          GenBankmRNA BI601514.1 - 2, 3 431 Chr. 3 - 181699575 - 181739998 [+] on Build GRCh38
          GenBankmRNA DA277798.1 - 1, 2 292 Chr. 3 - 181699612 - 181739778 [+] on Build GRCh38
          GenBankmRNA DA278757.1 - 1, 2 323 Chr. 3 - 181699577 - 181739805 [+] on Build GRCh38
          GenBankmRNA DA362437.1 - 1, 2 321 Chr. 3 - 181699575 - 181739729 [+] on Build GRCh38
          GenBankmRNA DA346811.1 - 2, 3 430 Chr. 3 - 181699577 - 181739706 [+] on Build GRCh38
          GenBankmRNA DA308672.1 - 2, 3 463 Chr. 3 - 181610363 - 181739693 [+] on Build GRCh38
          GenBankmRNA BI549819.1 - 1, 2 310 Chr. 3 - 181699589 - 181740014 [+] on Build GRCh38
          GenBankmRNA DB175439.1 - 1, 2 353 Chr. 3 - 181699546 - 181739816 [+] on Build GRCh38
          GenBankmRNA DA352078.1 - 1, 2 320 Chr. 3 - 181699575 - 181739818 [+] on Build GRCh38
          GenBankmRNA DA277074.1 - 2, 3 431 Chr. 3 - 181699575 - 181739726 [+] on Build GRCh38
          GenBankmRNA DA160000.1 - 1, 2 322 Chr. 3 - 181699575 - 181739815 [+] on Build GRCh38
          GenBankmRNA DA531399.1 - 1, 2 321 Chr. 3 - 181699576 - 181739876 [+] on Build GRCh38
          GenBankmRNA DA285311.1 - 2, 3 429 Chr. 3 - 181699575 - 181739689 [+] on Build GRCh38
          GenBankmRNA DA303463.1 - 2, 3 430 Chr. 3 - 181699577 - 181739737 [+] on Build GRCh38
          GenBankmRNA DA270443.1 - 2, 3 400 Chr. 3 - 181699604 - 181739755 [+] on Build GRCh38
          GenBankmRNA DA363633.1 - 1, 2 332 Chr. 3 - 181699575 - 181739821 [+] on Build GRCh38
          GenBankmRNA DA168440.1 - 1, 2 321 Chr. 3 - 181699575 - 181739847 [+] on Build GRCh38
          GenBankmRNA BM725940.1 - 1, 2 322 Chr. 3 - 181699573 - 181739882 [+] on Build GRCh38
          GenBankmRNA DA283659.1 - 2, 3 408 Chr. 3 - 181699596 - 181739753 [+] on Build GRCh38
          GenBankmRNA DA181278.1 - 1, 2 308 Chr. 3 - 181699587 - 181739839 [+] on Build GRCh38
          GenBankmRNA DA367963.1 - 2, 3 428 Chr. 3 - 181699575 - 181739710 [+] on Build GRCh38
          GenBankmRNA DA279363.1 - 1, 2 320 Chr. 3 - 181699575 - 181739844 [+] on Build GRCh38
          GenBankmRNA DA345402.1 - 2, 3 429 Chr. 3 - 181699575 - 181739714 [+] on Build GRCh38
          GenBankmRNA DA145889.1 - 1, 2 323 Chr. 3 - 181699575 - 181739823 [+] on Build GRCh38
          GenBankmRNA DA342800.1 - 2, 3 428 Chr. 3 - 181699575 - 181739728 [+] on Build GRCh38
          GenBankmRNA DA357231.1 - 2, 3 392 Chr. 3 - 181699612 - 181739753 [+] on Build GRCh38
          GenBankmRNA DA410724.1 - 2, 3 425 Chr. 3 - 181699580 - 181739615 [+] on Build GRCh38
          GenBankmRNA DA134835.1 - 1, 2 311 Chr. 3 - 181699585 - 181739838 [+] on Build GRCh38
          GenBankmRNA DA286657.1 - 2, 3 459 Chr. 3 - 181699546 - 181739688 [+] on Build GRCh38
          GenBankmRNA DB474842.1 - 2, 3 430 Chr. 3 - 181699575 - 181739664 [+] on Build GRCh38
          GenBankmRNA BF347925.1 - 1, 2 364 Chr. 3 - 181699529 - 181739824 [+] on Build GRCh38
          GenBankmRNA BI757585.1 - 1, 2 382 Chr. 3 - 181699515 - 181740032 [+] on Build GRCh38
          GenBankmRNA DA159447.1 - 2, 3 429 Chr. 3 - 181699575 - 181739721 [+] on Build GRCh38
          GenBankmRNA DA284362.1 - 1, 2 320 Chr. 3 - 181699575 - 181739749 [+] on Build GRCh38
          GenBankmRNA DA406068.1 - 2, 3 429 Chr. 3 - 181699576 - 181739736 [+] on Build GRCh38
          GenBankmRNA DA166593.1 - 2, 3 434 Chr. 3 - 181699577 - 181739690 [+] on Build GRCh38
          GenBankmRNA DA351387.1 - 2, 3 431 Chr. 3 - 181699575 - 181739739 [+] on Build GRCh38
          GenBankmRNA DB173817.1 - 1, 2 339 Chr. 3 - 181699559 - 181739811 [+] on Build GRCh38
          GenBankmRNA DA264513.1 - 1, 2 321 Chr. 3 - 181699575 - 181739815 [+] on Build GRCh38
          GenBankmRNA Z44703.1 - 288 Chr. - - [-] on Build
          GenBankmRNA DA273021.1 - 1, 2 321 Chr. 3 - 181699575 - 181739838 [+] on Build GRCh38
          GenBankmRNA BI916139.1 - 2, 3 476 Chr. 3 - 181699526 - 181739917 [+] on Build GRCh38
          GenBankmRNA DA406378.1 - 1, 2 390 Chr. 3 - 181699505 - 181739620 [+] on Build GRCh38
          GenBankmRNA BI668091.1 - 2, 3 428 Chr. 3 - 181699598 - 181739968 [+] on Build GRCh38
          GenBankmRNA DA298250.1 - 2, 3 392 Chr. 3 - 181699612 - 181739750 [+] on Build GRCh38
          GenBankmRNA HY126742.1 - 1, 2 298 Chr. 3 - 181699597 - 181739750 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0