Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BRCA1 SIRNA Assays ›
Gene Symbol:
BRCA1
Gene Name:
BRCA1, DNA repair associated
Gene Aliases:
BRCAI, BRCC1, BROVCA1, FANCS, IRIS, PNCA4, PPP1R53, PSCP, RNF53
Chromosome Location:
Chr.17: 43044295 - 43125483 on Build GRCh38
UniGene ID:
Hs.194143
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n264125
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BRCA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027676.1
1
115
GenBank mRNA
AB621825.1
-
1
156
AF005068.1
-
1
80
AK308084.1
-
1
115
DQ333386.1
-
1
121
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
double-strand break repair via homologous recombination
DNA double-strand break processing
DNA synthesis involved in DNA repair
strand displacement
DNA replication
postreplication repair
double-strand break repair
double-strand break repair via nonhomologous end joining
regulation of gene expression by genetic imprinting
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
regulation of transcription from RNA polymerase III promoter
fatty acid biosynthetic process
apoptotic process
cellular response to DNA damage stimulus
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
chromosome segregation
centrosome cycle
brain development
response to nutrient
intrinsic apoptotic signaling pathway in response to DNA damage
dosage compensation by inactivation of X chromosome
response to ionizing radiation
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
protein ubiquitination
protein sumoylation
androgen receptor signaling pathway
chromosome breakage
positive regulation of protein ubiquitination
G2 DNA damage checkpoint
response to estradiol
negative regulation of intracellular estrogen receptor signaling pathway
positive regulation of protein import into nucleus, translocation
positive regulation of histone acetylation
negative regulation of histone acetylation
regulation of cell proliferation
regulation of apoptotic process
chordate embryonic development
response to estrogen
regulation of DNA methylation
negative regulation of fatty acid biosynthetic process
positive regulation of DNA repair
positive regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of centriole replication
positive regulation of histone H3-K4 methylation
negative regulation of histone H3-K4 methylation
negative regulation of histone H3-K9 methylation
positive regulation of histone H3-K9 methylation
protein autoubiquitination
positive regulation of histone H4-K20 methylation
positive regulation of cell cycle arrest
cellular response to tumor necrosis factor
cellular response to indole-3-methanol
protein K6-linked ubiquitination
regulation of signal transduction by p53 class mediator
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of reactive oxygen species metabolic process
positive regulation of histone H3-K9 acetylation
positive regulation of histone H4-K16 acetylation
DNA binding
chromatin binding
damaged DNA binding
transcription coactivator activity
RNA binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
tubulin binding
ligase activity
enzyme binding
ubiquitin protein ligase binding
transcription regulatory region DNA binding
androgen receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-p5628:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0