Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMAD6 SIRNA Assays ›
Gene Symbol:
SMAD6
Gene Name:
SMAD family member 6
Gene Aliases:
AOVD2, HsT17432, MADH6, MADH7
Chromosome Location:
Chr.15: 66702110 - 66782000 on Build GRCh38
UniGene ID:
Hs.153863
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n264207
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMAD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027654.1
3, 4
1886
GenBank mRNA
AM909653.1
-
3, 4
963
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
outflow tract septum morphogenesis
mitral valve morphogenesis
pulmonary valve morphogenesis
ventricular septum development
transcription, DNA-templated
immune response
transforming growth factor beta receptor signaling pathway
zygotic specification of dorsal/ventral axis
negative regulation of cell proliferation
negative regulation of SMAD protein complex assembly
negative regulation of ossification
BMP signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of BMP signaling pathway
cell-substrate adhesion
response to laminar fluid shear stress
aorta development
negative regulation of apoptotic process
response to estrogen
fat cell differentiation
negative regulation of pathway-restricted SMAD protein phosphorylation
coronary vasculature development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
transforming growth factor beta receptor, inhibitory cytoplasmic mediator activity
ubiquitin protein ligase binding
type I transforming growth factor beta receptor binding
identical protein binding
transcription regulatory region DNA binding
metal ion binding
co-SMAD binding
I-SMAD binding
R-SMAD binding
type I activin receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0