siRNA Details
Gene Symbol: BZW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207067.1 | NP_001193996.1 | 9 | 1371 | ||
NM_001207069.1 | NP_001193998.1 | 9 | 1051 | |||
NM_001321688.1 | NP_001308617.1 | 9 | 951 | |||
NM_001321690.1 | NP_001308619.1 | 9 | 1065 | |||
NM_001321691.1 | NP_001308620.1 | 9 | 995 | |||
NM_001321693.1 | NP_001308622.1 | 8 | 818 | |||
NM_001321694.1 | NP_001308623.1 | 9 | 926 | |||
XM_017005393.1 | XP_016860882.1 | 6 | 664 | |||
GenBank mRNA | AK096489.1 | - | 8 | 928 | ||
AK223245.1 | - | 9 | 980 | |||
AK297227.1 | - | 9 | 1064 | |||
AK301511.1 | - | 9 | 1051 | |||
AL833518.1 | - | 9 | 1327 | |||
BC026303.1 | - | 9 | 980 | 4284614 | ||
D13630.1 | - | 9 | 996 |

More Information
Panther Classification:
Molecular Function -





