Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003090.2 | 6 | 2285 | |||
NR_103504.1 | 7 | 2627 | ||||
NR_103505.1 | 7 | 2665 | ||||
GenBank mRNA | BC022471.1 | - | 6 | 2180 |