Search
Search

GALNTL5| Gene Symbol: GALNTL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033169.1 | 1, 2 | 226 | |||
| XM_006715862.3 | XP_006715925.1 | 1, 2 | 197 | |||
| GenBank mRNA | AF440400.1 | - | 1, 2 | 161 | ||
| AK093091.1 | - | 1, 2 | 179 | |||
| AK315667.1 | - | 1, 2 | 185 | |||