Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289922.2 | NP_001276851.1 | 6 | 1690 | ||
XM_017028796.1 | XP_016884285.1 | 5 | 5127 | |||
GenBank mRNA | AK124962.1 | - | 6 | 1621 | ||
AK126430.1 | - | 4 | 2159 | |||
BC048207.1 | - | 4 | 628 | |||
BM051939.1 | - | 4 | 624 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM051456.1 | - | 4 | 624 | Chr. 22 - 49620423 - 49657538 [-] on Build GRCh38 |