Search
Search

DPY19L2P2| Gene Symbol: DPY19L2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003561.2 | 5 | 450 | |||
| NR_027768.1 | 3 | 745 | ||||
| GenBank mRNA | AK097711.1 | - | 3 | 673 | ||
| AK302046.1 | - | 4 | 658 | |||
| AL834175.1 | - | 3 | 498 | |||
| BC023575.2 | - | 4 | 847 | |||
| BC068601.1 | - | 3 | 556 | |||
| BC150309.1 | - | 3 | 206 | |||
| BX648212.1 | - | 4 | 727 | |||
| DB094748.1 | - | 4 | 349 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB068315.1 | - | 5 | 426 | Chr. 7 - 103275459 - 103280262 [-] on Build GRCh38 | |