Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR199A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029618.1 | 1 | 65 |
Gene Symbol: DNM3OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038397.2 | 1 | 256 | |||
GenBank mRNA | FJ623959.1 | - | 1 | 256 |