Search
Search

NMRK1| Gene Symbol: NMRK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_023352.1 | 3 | 444 | |||
| XM_006717162.1 | XP_006717225.1 | 3 | 472 | |||
| XM_011518817.1 | XP_011517119.1 | 2 | 297 | |||
| XM_011518818.1 | XP_011517120.1 | 2 | 819 | |||
| XM_011518819.2 | XP_011517121.1 | 2 | 819 | |||
| XM_017014868.1 | XP_016870357.1 | 4 | 793 | |||
| XM_017014869.1 | XP_016870358.1 | 3 | 1100 | |||
| XM_017014871.1 | XP_016870360.1 | 3 | 472 | |||
| XM_017014872.1 | XP_016870361.1 | 4 | 793 | |||
| XM_017014873.1 | XP_016870362.1 | 4 | 764 | |||
| GenBank mRNA | AK097144.1 | - | 3 | 417 | ||