Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS31P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002816.2 | 9 | 2726 | |||
NR_051963.1 | 9 | 2011 | ||||
NR_051964.1 | 7 | 1596 | ||||
NR_051965.1 | 8 | 1692 | ||||
GenBank mRNA | AK027536.1 | - | 9 | 2726 | ||
BX648914.1 | - | 9 | 2001 |