Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002959.1 | 1 | 98 | |||
GenBank mRNA | AJ609471.1 | - | 1 | 98 |
Gene Symbol: TAF1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128061.1 | - | 1 | 2349 |