Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM41C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027055.1 | 3 | 1655 |
Gene Symbol: LOC101927506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014134.1 | XP_016869623.1 | 4 | 2755 |
Gene Symbol: LOC102723872 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515681.1 | XP_011513983.1 | 5 | 1127 |
Gene Symbol: RPL23AP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL365511.1 | - | 4 | 807 |