siRNA Details
Gene Symbol: AGPAT4-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024277.1 | 1 | 139 | |||
GenBank mRNA | AK026765.1 | - | 1 | 147 |

Gene Symbol: AGPAT4-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024277.1 | 1 | 139 | |||
GenBank mRNA | AK026765.1 | - | 1 | 147 |