Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026857.1 | 2 | 2268 | |||
GenBank mRNA | AK027350.1 | - | 1 | 132 | ||
AK056555.1 | - | 2 | 2250 | |||
AK075238.1 | - | 1 | 231 | |||
AK096609.1 | - | 1 | 1176 | |||
BC110822.1 | - | 1 | 277 |