Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYO15B SIRNA Assays ›
Gene Symbol:
MYO15B
Gene Name:
myosin XVB
Gene Aliases:
MYO15BP
Chromosome Location:
Chr.17: 75587545 - 75626850 on Build GRCh38
UniGene ID:
Hs.390817
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n268139
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYO15B
Interrogated Sequence
Translated Protein
RNADB ID
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001309242.1
NP_001296171.1
63
9248
XM_017025120.1
XP_016880609.1
65
8955
XM_017025121.1
XP_016880610.1
65
8940
XM_017025122.1
XP_016880611.1
65
8919
XM_017025123.1
XP_016880612.1
65
8891
XM_017025124.1
XP_016880613.1
64
8829
XM_017025125.1
XP_016880614.1
63
8678
XM_017025126.1
XP_016880615.1
60
8420
XM_017025127.1
XP_016880616.1
63
8263
XM_017025128.1
XP_016880617.1
64
8009
XM_017025129.1
XP_016880618.1
63
7241
XM_017025130.1
XP_016880619.1
62
7197
XM_017025131.1
XP_016880620.1
61
7636
XM_017025132.1
XP_016880621.1
60
7577
XM_017025133.1
XP_016880622.1
58
7102
XM_017025134.1
XP_016880623.1
59
7463
XM_017025135.1
XP_016880624.1
53
6119
XM_017025136.1
XP_016880625.1
53
6468
XM_017025137.1
XP_016880626.1
50
6013
XM_017025138.1
XP_016880627.1
45
5509
XM_017025139.1
XP_016880628.1
46
5310
XM_017025140.1
XP_016880629.1
43
5123
XM_017025143.1
XP_016880632.1
38
4400
XM_017025144.1
XP_016880633.1
59
8883
GenBank mRNA
AK026339.1
-
15
1563
AK096869.1
-
15
1702
AL713730.1
-
14
1457
BC027875.1
-
LIT3316
MYO15BP
15
1952
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
HY104121.1
-
2
286
Chr. 17 - 75626092 - 75626716 [+] on Build GRCh38
GenBankmRNA
BX110022.1
-
3
419
Chr. 17 - 75625855 - 75626849 [+] on Build GRCh38
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0