Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DHRS4-AS1 SIRNA Assays ›
          Gene Symbol:
          DHRS4-AS1
          Gene Name:
          DHRS4 antisense RNA 1
          Gene Aliases:
          AS1DHRS4, C14orf167, DHRS4AS1, PRO1488
          Chromosome Location:
          Chr.14: 23938731 - 23955112 on Build GRCh38
          UniGene ID:
          Hs.569361
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n268147

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DHRS4-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_023921.2 2 820
          NR_023922.2 2 561
          NR_023923.2 3 529
          NR_023924.2 2 620
          GenBank mRNA BC080582.1 - 2 547
          BF206895.1 - 2 539
          BX247982.1 - 2 290
          BX447332.2 - 2 744
          BX482101.1 - 2 562
          BX647964.1 - 3 499
          DB254318.1 - 3 515
          HQ845197.1 - 3 515
          HQ845198.1 - 3 515
          HQ845199.1 - 2 600
          JQ979294.1 - 3 515
          KJ545484.1 - 2 456
          KJ574270.1 - 2 247
          KJ574271.1 - 2 380
          KJ574272.1 - 2 379
          KJ574273.1 - 2 539
          KJ574274.1 - 2 583
          KJ599455.1 - 2 313
          KJ599456.1 - 2 570
          KJ599457.1 - 2 272
          KJ599458.1 - 1 66
          KJ599459.1 - 3 488
          KJ599460.1 - 2 614
          KJ599461.1 - 3 498
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA085243.1 - 2 560 Chr. 14 - 23940945 - 23954137 [-] on Build GRCh38
          GenBankmRNA DA765281.1 - 3 480 Chr. 14 - 23940868 - 23954148 [-] on Build GRCh38
          GenBankmRNA BX461600.2 - 2 291 Chr. 14 - 23940338 - 23953668 [-] on Build GRCh38
          GenBankmRNA BE408218.1 - 2 523 Chr. 14 - 23940818 - 23954102 [-] on Build GRCh38
          GenBankmRNA BG257690.1 - 2 585 Chr. 14 - 23940706 - 23954164 [-] on Build GRCh38
          GenBankmRNA DA565385.1 - 2 469 Chr. 14 - 23940867 - 23954035 [-] on Build GRCh38
          GenBankmRNA CN277983.1 - 3 493 Chr. 14 - 23940714 - 23954156 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-fq9zm:80/100.66.76.89:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0