Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DHRS4-AS1 SIRNA Assays ›
          Gene Symbol:
          DHRS4-AS1
          Gene Name:
          DHRS4 antisense RNA 1
          Gene Aliases:
          AS1DHRS4, C14orf167, DHRS4AS1, PRO1488
          Chromosome Location:
          Chr.14: 23938731 - 23955112 on Build GRCh38
          UniGene ID:
          Hs.569361
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n268149

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DHRS4-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_023921.2 2 795
          NR_023922.2 2 536
          NR_023923.2 3 504
          NR_023924.2 2 595
          GenBank mRNA BC080582.1 - 2 522
          BF206895.1 - 2 514
          BX247982.1 - 2 265
          BX447332.2 - 2 719
          BX482101.1 - 2 537
          DB254318.1 - 3 490
          HQ845197.1 - 3 490
          HQ845198.1 - 3 490
          HQ845199.1 - 2 575
          JQ979294.1 - 3 490
          KJ545484.1 - 2 431
          KJ574270.1 - 2 222
          KJ574271.1 - 2 355
          KJ574272.1 - 2 354
          KJ574273.1 - 2 514
          KJ599456.1 - 2 545
          KJ599457.1 - 2 247
          KJ599458.1 - 1 41
          KJ599460.1 - 2 589
          KJ599461.1 - 3 473
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB007427.1 - 2 533 Chr. 14 - 23940966 - 23954135 [-] on Build GRCh38
          GenBankmRNA AL692221.1 - 2 555 Chr. 14 - 23940961 - 23954156 [-] on Build GRCh38
          GenBankmRNA DA085243.1 - 2 535 Chr. 14 - 23940945 - 23954137 [-] on Build GRCh38
          GenBankmRNA DA765281.1 - 3 455 Chr. 14 - 23940868 - 23954148 [-] on Build GRCh38
          GenBankmRNA BX461600.2 - 2 266 Chr. 14 - 23940338 - 23953668 [-] on Build GRCh38
          GenBankmRNA BE408218.1 - 2 498 Chr. 14 - 23940818 - 23954102 [-] on Build GRCh38
          GenBankmRNA BG257690.1 - 2 560 Chr. 14 - 23940706 - 23954164 [-] on Build GRCh38
          GenBankmRNA DA565385.1 - 2 444 Chr. 14 - 23940867 - 23954035 [-] on Build GRCh38
          GenBankmRNA DA442325.1 - 2 564 Chr. 14 - 23940978 - 23954166 [-] on Build GRCh38
          GenBankmRNA DB482758.1 - 3 465 Chr. 14 - 23940961 - 23954158 [-] on Build GRCh38
          GenBankmRNA CN277983.1 - 3 468 Chr. 14 - 23940714 - 23954156 [-] on Build GRCh38
          GenBankmRNA BX404846.2 - 2 719 Chr. 14 - 23940714 - 23954122 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0