Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASMTL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026710.1 | 4 | 969 | |||
NR_026711.1 | 4 | 969 | ||||
GenBank mRNA | AF193043.2 | - | 3 | 332 | ||
AK023392.1 | - | 4 | 402 | |||
BC093994.2 | - | 3 | 312 | |||
BC143527.1 | - | 3 | 346 |
Gene Symbol: ASMTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU688748.1 | - | 2 | 404 |